Skip to Content
Merck
All Photos(1)

Documents

V0030000

Valine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Valine, (S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein:
1721136
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

valine

manufacturer/tradename

EDQM

mp

295-300 °C (subl.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1

InChI key

KZSNJWFQEVHDMF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Valine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Leclercq
Poultry science, 77(1), 118-123 (1998-02-20)
Lysine exerts specific effects on body composition at dietary levels higher than that required for maximum growth rate, which also results in an improved feed conversion ratio (FCR). As a consequence of these differences, it becomes apparent that there is
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
Ijin Joo et al.
Radiology, 272(2), 417-426 (2014-04-05)
To evaluate the diagnostic value of intravoxel incoherent motion (IVIM) diffusion-weighted (DW) magnetic resonance (MR) imaging in the quantitative assessment of the therapeutic efficacy of a vascular disrupting agent (VDA) (CKD-516) in rabbit VX2 liver tumors. The institutional animal care
Growth inhibition as a consequence of antagonism between related amino acids: effect of valine in Escherichia coli K-12.
M De Felice et al.
Microbiological reviews, 43(1), 42-58 (1979-03-01)
Xin Zhang et al.
Hypertension (Dallas, Tex. : 1979), 64(1), 87-93 (2014-04-23)
Renovascular hypertension alters cardiac structure and function. Autophagy is activated during left ventricular hypertrophy and linked to adverse cardiac function. The angiotensin II receptor blocker, valsartan, lowers blood pressure and is cardioprotective, but whether it modulates autophagy in the myocardium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service