Skip to Content
Merck
All Photos(1)

Key Documents

T1610000

RRR-α-Tocopheryl hydrogen succinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-α-Tocopherol succinate, Vitamin E succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H54O5
CAS Number:
Molecular Weight:
530.78
Beilstein:
4038233
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

tocopherol

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)CCC(O)=O)c(C)c(C)c2O1

InChI

1S/C33H54O5/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-33(8)21-19-28-27(7)31(25(5)26(6)32(28)38-33)37-30(36)18-17-29(34)35/h22-24H,9-21H2,1-8H3,(H,34,35)/t23-,24-,33-/m1/s1

InChI key

IELOKBJPULMYRW-NJQVLOCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

RRR-α-Tocopheryl hydrogen succinate, the ester of RRR-α-tocopherol, is a primary source of vitamin E for consumption as a component of the diet.[1]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

RRR-α-Tocopheryl hydrogen succinate for peak identification may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by gas chromatography (GC).[2]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wild Mammals in Captivity: Principles and Techniques for Zoo Management, Second Edition
Wild Mammals in Captivity (2010)
RRR-?-Tocopheryl hydrogen succinate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3808-3810 (2017)
Pengfei Xu et al.
Biomaterials, 35(26), 7574-7587 (2014-06-06)
Metastasis is one of the major obstacles for the successful therapy of breast cancer. Although increased candidate drugs targeting cancer metastasis are tested, their clinical translation is limited by either serve toxicity or low efficacy. In present work, a nano-drug
Sindhuri Maddineni et al.
AAPS PharmSciTech, 16(2), 444-454 (2014-11-02)
The objective of the present study was to investigate the effects of processing variables and formulation factors on the characteristics of hot-melt extrudates containing a copolymer (Kollidon® VA 64). Nifedipine was used as a model drug in all of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service