Skip to Content
Merck
All Photos(1)

Key Documents

T1423000

Ticlopidine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClNS · HCl
CAS Number:
Molecular Weight:
300.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ticlopidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.Clc1ccccc1CN2CCc3sccc3C2

InChI

1S/C14H14ClNS.ClH/c15-13-4-2-1-3-11(13)9-16-7-5-14-12(10-16)6-8-17-14;/h1-4,6,8H,5,7,9-10H2;1H

InChI key

MTKNGOHFNXIVOS-UHFFFAOYSA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ticlopidine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morten Lamberts et al.
Circulation, 129(15), 1577-1585 (2014-01-29)
The optimal long-term antithrombotic treatment of patients with coexisting atrial fibrillation and stable coronary artery disease is unresolved, and commonly, a single antiplatelet agent is added to oral anticoagulation. We investigated the effectiveness and safety of adding antiplatelet therapy to
Stephen D Wiviott et al.
Lancet (London, England), 382(9892), 605-613 (2013-08-21)
Treatment with prasugrel and aspirin improves outcomes compared with clopidogrel and aspirin for patients with acute coronary syndrome who have had angiography and percutaneous coronary intervention; however, no clear benefit has been shown for patients managed first with drugs only.
Willibald Hochholzer et al.
Journal of the American College of Cardiology, 64(4), 361-368 (2014-07-26)
The degree of antiplatelet response to clopidogrel has been associated with clinical outcomes. Studies have investigated whether adjustment of antiplatelet therapies based on a single platelet function test is beneficial. The aim of the study was to test the stability
A Tornio et al.
Clinical pharmacology and therapeutics, 96(4), 498-507 (2014-06-28)
Cerivastatin and repaglinide are substrates of cytochrome P450 (CYP)2C8, CYP3A4, and organic anion-transporting polypeptide (OATP)1B1. A recent study revealed an increased risk of rhabdomyolysis in patients using cerivastatin with clopidogrel, warranting further studies on clopidogrel interactions. In healthy volunteers, repaglinide
Colin P Derdeyn et al.
Lancet (London, England), 383(9914), 333-341 (2013-10-31)
Early results of the Stenting and Aggressive Medical Management for Preventing Recurrent stroke in Intracranial Stenosis trial showed that, by 30 days, 33 (14·7%) of 224 patients in the stenting group and 13 (5·8%) of 227 patients in the medical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service