Skip to Content
Merck
All Photos(1)

Documents

T0100000

Testosterone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17β-Hydroxy-3-oxo-4-androstene, 17β-Hydroxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one, trans-Testosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
Beilstein:
1915399
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

testosterone

manufacturer/tradename

EDQM

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1

InChI key

MUMGGOZAMZWBJJ-DYKIIFRCSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testosterone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Testosterone secreted by the testis is converted to dihydrotestosterone in the target tissues where it appears to mediate many of the biological actions of testosterone. Androgens direct the development of the male phenotype during embryogenesis and at puberty.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Zimmerman et al.
Human reproduction update, 20(1), 76-105 (2013-10-02)
BACKGROUND; Combined oral contraceptives (COCs) reduce levels of androgen, especially testosterone (T), by inhibiting ovarian and adrenal androgen synthesis and by increasing levels of sex hormone-binding globulin (SHBG). Although this suppressive effect has been investigated by numerous studies over many
Marie Sinclair et al.
Transplantation, 98(7), 788-792 (2014-06-12)
Low pretransplant serum testosterone has recently been associated with increased mortality in men awaiting liver transplantation, but the potential impact on rejection has not yet been investigated. Pretransplantation serum testosterone, SHBG, and other variables were collected on 190 consecutive men
David J Handelsman
The Journal of clinical endocrinology and metabolism, 91(5), 1646-1653 (2006-02-16)
The objective of the study was to review the rationale underlying the banning of human chorionic gonadotropin (hCG) and estrogen blockers (antiestrogens, specific estrogen receptor modulators, aromatase inhibitors) in sports for male and female athletes in the light of gender
James R Bell et al.
American journal of physiology. Heart and circulatory physiology, 306(9), H1265-H1274 (2014-03-13)
Estrogen in females is conventionally considered a cardioprotective influence, but a role for estrogen in male cardioprotection has yet to be defined. Estrogen biosynthesis from testosterone is regulated by aromatase. Aromatase has recently been shown to be expressed in the
Andrea C Lo et al.
International journal of radiation oncology, biology, physics, 88(1), 87-93 (2013-12-18)
To determine (1) the prognostic utility of prostate-specific antigen (PSA) concentration at 45 to 60 months (48mPSA) after low-dose-rate prostate brachytherapy (LDR-PB); (2) the predictors of 48mPSA; and (3) the prognostic utility of directional trends between PSA levels at 24

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service