Skip to Content
Merck
All Photos(1)

Key Documents

SMB00974

Sigma-Aldrich

Albendazole sulfone

≥95% (HPLC)

Synonym(s):

[5-(Propane-1-sulfonyl)-1H-benzoimidazol-2-yl]-carbamic acid methyl ester, Methyl [5-(propylsulfonyl)-1H-benzimidazol-2-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O4S
CAS Number:
Molecular Weight:
297.33
Beilstein:
8394880
MDL number:
UNSPSC Code:
51451806
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

CCCS(=O)(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O4S/c1-3-6-20(17,18)8-4-5-9-10(7-8)14-11(13-9)15-12(16)19-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

CLSJYOLYMZNKJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Albendazole sulfone is a major inactive metabolite of anti-parasitic agent albendazole.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,spleen,male reproductive organs,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andi Dian Permana et al.
Journal of pharmaceutical and biomedical analysis, 170, 243-253 (2019-04-05)
A sensitive and relatively fast, cost-effective high-performance liquid chromatographic method coupled with mass spectrometer (HPLC-MS) is herein reported for the first time for a simultaneous quantification of plasma and organs concentration of three therapeutic agents that are widely used in
Jessica D Schulz et al.
Antimicrobial agents and chemotherapy, 63(4) (2019-02-13)
Albendazole is an effective anthelmintic intensively used for decades. However, profound pharmacokinetic (PK) characterization is missing in children, the population mostly affected by helminth infections. Blood microsampling would facilitate PK studies in pediatric populations but has not been applied to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service