Skip to Content
Merck
All Photos(3)

Key Documents

S6172

Sigma-Aldrich

Sodium persulfate

BioXtra, ≥99%

Synonym(s):

Sodium peroxodisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O8
CAS Number:
Molecular Weight:
238.10
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99%
solubility:
H2O: 1 M at 20 °C, clear, colorless

product line

BioXtra

Quality Level

Assay

≥99%

reaction suitability

reagent type: oxidant

impurities

<0.0005% Phosphorus (P)
<0.1% Insoluble matter

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.02%
Mg: <0.001%
Pb: <0.001%
Zn: <0.0005%

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

CHQMHPLRPQMAMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Electrochemical treatment of organic pollutants in landfill leachate using a three-dimensional electrode system.</strong>: This study explores the electrochemical treatment of landfill leachate using a three-dimensional electrode system. Sodium persulfate is used as an oxidizing agent to degrade organic pollutants effectively, providing a potential method for waste management and environmental protection (Yu et al., 2020).</li>
<li><strong>The Box-Benkhen experimental design for the optimization of the electrocatalytic treatment of wastewaters with high concentrations of phenol and organic matter.</strong>: This paper discusses the optimization of electrocatalytic treatment processes for wastewater containing high levels of phenol and organic matter using sodium persulfate. The study provides valuable insights for improving wastewater treatment efficiency (GilPavas et al., 2009).</li>
<li><strong>Reaction of pectin and glycidyl methacrylate and ulterior formation of free films by reticulation.</strong>: This research involves the chemical modification of pectin with glycidyl methacrylate followed by cross-linking using sodium persulfate, leading to the formation of free-standing films. These films have potential applications in pharmaceuticals and food packaging (Maior et al., 2008).</li>
</ul>

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Hsien Yen et al.
Journal of hazardous materials, 186(2-3), 2097-2102 (2011-01-25)
In this study, batch experiments were conducted to evaluate the feasibility of petroleum-hydrocarbon contaminated soil remediation using persulfate oxidation. Various controlling factors including different persulfate and ferrous ion concentrations, different oxidants (persulfate, hydrogen peroxide, and permanganate), and different contaminants (diesel
José Fenoll et al.
Journal of hazardous materials, 244-245, 370-379 (2012-12-29)
The photocatalytic degradation of sixteen substituted phenylurea herbicides (PUHs) in pure water has been studied using zinc oxide (ZnO) and titanium dioxide (TiO(2)) as photocatalyst under artificial light irradiation. Photocatalytic experiments showed that the addition of these chalcogenide oxides in
Xiaodan Zhao et al.
Journal of the American Chemical Society, 132(16), 5837-5844 (2010-04-03)
By palladium catalysis, the C-H bond functionalization of O-phenylcarbamates with simple arenes has been achieved using sodium persulfate (Na(2)S(2)O(8)), an inexpensive, easy-to-handle, and environmentally friendly oxidant. This oxidative cross-coupling involves two aromatic C-H bonds undergoing concomitant oxidation to furnish a
Chenju Liang et al.
Chemosphere, 70(3), 426-435 (2007-08-19)
In situ chemical oxidation with persulfate anion (S2O82*) is a viable technique for remediation of groundwater contaminants such as trichloroethylene (TCE). An accelerated reaction using S2O82* to destroy TCE can be achieved via chemical activation with ferrous ion to generate
N Sabri et al.
The Science of the total environment, 427-428, 382-389 (2012-05-15)
The objective of this work was to evaluate the removal of ibuprofen (IBP) using the oxidants hydrogen peroxide (H(2)O(2)) and sodium persulfate (Na(2)S(2)O(8)). The ability of magnetite (Fe(3)O(4)) to activate persulfate (PS) and H(2)O(2) for the oxidation of IBP at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service