Skip to Content
Merck
All Photos(1)

Key Documents

S0150000

Salbutamol sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Salbutamol hemisulfate salt, α1-[(tert-Butylamino)methyl]-4-hydroxy-1,3-benzenedimethanol hemisulfate salt, Albuterol sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NO3 · 0.5H2SO4
CAS Number:
Molecular Weight:
288.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

albuterol, salbutamol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OS(O)(=O)=O.CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2

InChI

1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4)

InChI key

BNPSSFBOAGDEEL-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salbutamol sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Padhukasahasram et al.
The pharmacogenomics journal, 14(4), 365-371 (2014-01-15)
Inhaled short-acting beta-agonist (SABA) medication is commonly used in asthma patients to rapidly reverse airway obstruction and improve acute symptoms. We performed a genome-wide association study of SABA medication response using gene-based association tests. A linear mixed model approach was
Ashley Woodcock et al.
Chest, 144(4), 1222-1229 (2013-07-13)
The combination of fluticasone furoate (FF), a novel inhaled corticosteroid (ICS), and vilanterol (VI), a long-acting β2 agonist, is under development as a once-daily treatment of asthma and COPD. The aim of this study was to compare the efficacy of
Huan Wang et al.
Biosensors & bioelectronics, 49, 14-19 (2013-05-28)
A multiplexed electrochemical biosensor has been developed for fast and sensitive detection of ractopamine (RAC), salbutamol (SAL) and clenbuterol (CLB) based on reduced graphene oxide (rGO) and silver-palladium alloy nanoparticles (AgPd NPs). In this paper, rGO with high conductivity was
Clinical Practice. Mild asthma.
Elisabeth H Bel
The New England journal of medicine, 369(6), 549-557 (2013-08-09)
Abebaw M Yohannes et al.
International journal of chronic obstructive pulmonary disease, 8, 117-125 (2013-03-22)
Tiotropium bromide is an anticholinergic agent that has gained worldwide acceptance as a first-line, once daily maintenance therapy for patients with moderate-to-severe chronic obstructive pulmonary disease. The purpose of this review is to synthesize the evidence base in the past

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service