Skip to Content
Merck
All Photos(2)

Key Documents

RDD038

Sigma-Aldrich

Sodium phosphate dibasic

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

product line

BioReagent
Redi-Dri

Quality Level

Assay

≥99.0%

form

powder or crystals

quality

free-flowing

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

pH

8.9-9.2 (25 °C, 50 g/L in water)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: soluble 100 mg/mL

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium phosphate dibasic is a component in many biological buffers used in biochemical, biomedical, and molecular biological research. It is also known as disodium hydrogen phosphate (Na2HPO4), it is an inorganic salt that is highly soluble in water. It serves as a versatile buffering agent in research with a pH of 8.9 - 9.2. It is essential for making several phosphate buffers including PBS, utilized for cell culture, immunohistochemistry, Western blot, ELISA, and other applications where a stable pH is critical.

Application

Sodium phosphate dibasic may be used for phosphate buffer and Phosphate Buffered Saline (PBS) that is commonly utilized for cell culture, immunohistochemistry, western blot, and other biochemical applications where a stable pH is crucial. Including protein purification and enzymatic assays.

Features and Benefits

  • Qualified for use in cell culture; suitable for insect cell culture
  • Redi-Dri packaging controls moisture for hygroscopic chemicals that commonly clump
  • Anhydrous powder remains free-flowing and easy to use
  • Safer to handle and weigh out for routine use in lab
  • Eliminates clumps, maintaining product moisture, quality, and attributes

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolating lipid droplets from multiple species
Ding Y, et al.
Nature Protocols, 8(1), 43-43 (2013)
Rapid and accurate analyses of silicon and phosphorus in plants using a portable X-ray fluorescence spectrometer
Reidinger S, et al.
The New phytologist, 195(3), 699-706 (2012)
Decellularization methods of porcine kidneys for whole organ engineering using a high-throughput system
Sullivan D C, et al.
Biomaterials, 33(31), 7756-7764 (2012)
Evaluation of Burkholderia cepacia complex bacteria pathogenicity using Caenorhabditis elegans
Tedesco P, et al.
Bio-protocol, 6(20) (2016)
Yan Li et al.
Structure (London, England : 1993), 26(4), 555-564 (2018-03-13)
Zika virus (ZIKV) infection has become a global public health concern. The viral NS2B-NS3 protease is an attractive antiviral target because of its role in maturation of viral non-structural proteins. Substrate-derived protease inhibitors have been investigated, but it remains challenging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service