Skip to Content
Merck
All Photos(1)

Key Documents

R1000000

Rifamycin sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Rifamycin SV sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H46NNaO12
CAS Number:
Molecular Weight:
719.75
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rifamycin, rifampicin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)cc([O-])c4c3C2=O

InChI

1S/C37H47NO12.Na/c1-16-11-10-12-17(2)36(46)38-23-15-24(40)26-27(32(23)44)31(43)21(6)34-28(26)35(45)37(8,50-34)48-14-13-25(47-9)18(3)33(49-22(7)39)20(5)30(42)19(4)29(16)41;/h10-16,18-20,25,29-30,33,40-44H,1-9H3,(H,38,46);/q;+1/p-1/b11-10+,14-13+,17-12-;/t16-,18+,19+,20+,25-,29-,30+,33+,37-;/m0./s1

InChI key

YVOFSHPIJOYKSH-NLYBMVFSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifamycin sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucy A Woods et al.
Nature chemical biology, 7(10), 730-739 (2011-08-30)
Although small molecules that modulate amyloid formation in vitro have been identified, significant challenges remain in determining precisely how these species act. Here we describe the identification of rifamycin SV as a potent inhibitor of β(2) microglobulin (β(2)m) fibrillogenesis when
Marta Calatayud et al.
Chemical research in toxicology, 25(2), 446-453 (2012-01-05)
Inorganic arsenic (iAs) [As(III)+As(V)] is a drinking water contaminant, and human exposure to these arsenic species has been linked with a wide range of health effects. The main path of exposure is the oral route, and the intestinal epithelium is
E Vigliaroli et al.
Minerva stomatologica, 59(9), 507-517 (2010-10-14)
Actinomycosis is an uncommon chronic granulomatous infection that cause formation of abscesses and cutaneous fistula. In mandibular actinomycosis the alveolar bone and mandibular body are usually not involved and the pathogenetic mechanisms of the actinomycotic infiltration is unknown. The patients
Wen-Tao Peng et al.
Acta biochimica et biophysica Sinica, 38(12), 821-830 (2006-12-08)
The structural gene for glutamine synthetase, glnA, from Amycolatopsis mediterranei U32 was cloned via screening a genomic library using the analog gene from Streptomyces coelicolor. The clone was functionally verified by complementing for glutamine requirement of an Escherichia coli glnA
[Ornithinimicrobium pekingense ocular infection].
E Borsali et al.
Medecine et maladies infectieuses, 41(6), 345-346 (2011-03-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service