Skip to Content
Merck
All Photos(1)

Key Documents

R0145000

Ramipril

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tritace, [2S,3aS,6aS]-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N2O5
CAS Number:
Molecular Weight:
416.51
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ramipril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(N1[C@](CCC2)([H])[C@]2([H])C[C@H]1C(O)=O)[C@H](C)N[C@H](C(OCC)=O)CCC3=CC=CC=C3

InChI

1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17-,18-,19-,20-/m0/s1

InChI key

HDACQVRGBOVJII-JBDAPHQKSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ramipril EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina De Ciuceis et al.
Hypertension (Dallas, Tex. : 1979), 64(4), 717-724 (2014-07-02)
Structural alterations of subcutaneous small-resistance arteries are associated with a worse clinical prognosis in hypertension and non-insulin-dependent diabetes mellitus. The effects of the direct renin inhibitor aliskiren on microvascular structure were never previously evaluated. Therefore, we investigated the effects of
April D Vuong et al.
The Annals of pharmacotherapy, 37(3), 412-419 (2003-03-18)
To evaluate the effectiveness of ramipril in the prevention and treatment of cardiovascular disease and determine its need for inclusion on a formulary. A MEDLINE and PubMed database search was conducted (1987-May 2002). Only journals written in the English language
Antonios Douros et al.
American journal of hypertension, 26(9), 1070-1075 (2013-06-12)
Ramipril, an inhibitor of the angiotensin-converting enzyme (ACEI), is a drug commonly used in the therapy of hypertension. ACEI-induced hepatotoxicity is rare, and most of the reported cases are associated with captopril. Here, we present the first case of ramipril-induced
W Linz et al.
Zeitschrift fur Kardiologie, 83 Suppl 4, 53-56 (1994-01-01)
ACE inhibitors induce an increase in kinin levels with subsequent release of nitric oxide (NO) and prostacyclin, as shown in cultured endothelial cells and isolated rat hearts. Isolated perfused working rat hearts continuously release kinins and prostacyclin. During ischemia after
Gregory T Warner et al.
Drugs, 62(9), 1381-1405 (2002-06-22)
Ramipril, an angiotensin-converting enzyme (ACE) inhibitor, is a prodrug which is rapidly hydrolysed after absorption to the active metabolite ramiprilat. Earlier trials have shown that ACE inhibitors, when given to patients with low ejection fractions, have reduced the relative risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service