Skip to Content
Merck
All Photos(1)

Key Documents

PHR1914

Supelco

2-Ethylhexanoic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Ethylhexanoic acid, 2-Ethylcaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CO2H
CAS Number:
Molecular Weight:
144.21
Beilstein:
1750468
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

vapor density

4.98 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
10 mmHg ( 115 °C)

API family

2-ethylhexanoic acid

form

liquid

CofA

current certificate can be downloaded

autoignition temp.

699 °F

expl. lim.

1.04 %, 135 °F
8.64 %, 188 °F

packaging

ampule of 3x1.2 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.425 (lit.)

bp

228 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

storage temp.

2-30°C

SMILES string

CCCCC(CC)C(O)=O

InChI

1S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

OBETXYAYXDNJHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB0955 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria C de Leeuw et al.
Toxicology letters, 303, 28-37 (2018-12-24)
In vitro assays to assess developmental neurotoxicity of chemicals are highly desirable. The murine neural embryonic stem cell test (ESTn) can mimic parts of early differentiation of embryonic brain and may therefore be useful for this purpose. The aim of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service