Skip to Content
Merck
All Photos(1)

Key Documents

PHR1431

Supelco

Clopidogrel Bisulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate, CLP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO2S · H2SO4
CAS Number:
Molecular Weight:
419.90
Beilstein:
9967887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001333
traceable to USP 1140430

API family

clopidogrel

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.COC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl

InChI

1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/t15-;/m0./s1

InChI key

FDEODCTUSIWGLK-RSAXXLAASA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Clopidogrel Bisulfate is a novel antiplatelet agent which is chemically related to ticlopidine. It acts against ischaemic stroke, myocardial infraction as well as vascular disease. It is reported to have exhibited clinical efficacy greater than that of aspirin, during large phase III trials.

Application

Clopidogrel Bisulfate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography, spectrophotometric and spectrodensitometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3034 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric and spectrodensitometric determination of Clopidogrel Bisulfate with kinetic study of its alkaline degradation.
Zaazaa HE, et al.
Talanta, 78(3), 874-884 (2009)
Stability-indicating liquid chromatography for determination of clopidogrel bisulfate in tablets: Application to content uniformity testing.
Alarfaj NA
Journal of Saudi Chemical Society, 16(1), 23-30 (2012)
Concurrent estimation of clopidogrel bisulfate and aspirin in tablets by validated RP-HPLC method.
Shrivastava PK, et al.
Indian Journal of Pharmaceutical Sciences, 70(5), 667-667 (2008)
A validated stability-indicating normal phase LC method for clopidogrel bisulfate and its impurities in bulk drug and pharmaceutical dosage form.
Rao DD, et al.
Journal of Pharmaceutical and Biomedical Analysis, 52(1), 160-165 (2010)
Xinmeng Chen et al.
Biopharmaceutics & drug disposition, 36(1), 64-68 (2014-09-27)
Erigeron breviscapus (Vant.) Hand-Mazz, a traditional Chinese medicine, is often co-prescribed with clopidogrel for the treatment of ischemic vascular diseases. Scutellarin is the representative bioactive flavonoid isolated from this herb. The aim of this study was to explore the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service