Skip to Content
Merck
All Photos(1)

Key Documents

PHR1376

Supelco

Loratadine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Loratadine, 4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene-1-piperidinecarboxylic acid ethyl ester, Loratidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N2O2Cl
CAS Number:
Molecular Weight:
382.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 874
traceable to Ph. Eur. Y0000688
traceable to USP 1370270

API family

loratadine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(=O)N1CC\C(CC1)=C2/c3ccc(Cl)cc3CCc4cccnc24

InChI

1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3

InChI key

JCCNYMKQOSZNPW-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Loratadine belongs to the class of H1-antihistamine drugs, with non-sedating properties, and is widely used in the treatment of allergies.

Application

Loratadine may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Non-sedating histamine H1-receptor antagonist.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3785 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of loratadine in pharmaceuticals by a spectrophotometric method
Pavalanche G, et al.
Ovidius University Annals of Chemistry, 26(1), 27-31 (2015)
J A Hey et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 29 Suppl 3, 197-199 (1999-08-12)
The extremely low reporting rate of cardiovascular adverse events for loratadine, the possible preferential use of loratadine in patients with pre-existing cardiovascular disorders, and the impressive lack of cardiovascular effects at extremely high concentrations in clinical and preclinical studies demonstrate
S P Clissold et al.
Drugs, 37(1), 42-57 (1989-01-01)
Loratadine is a long acting antihistamine which has a high selectivity for peripheral histamine H1-receptors and lacks the central nervous system depressant effects often associated with some of the older antihistamines. Results from controlled clinical trials have shown that loratadine
H A Barenholtz et al.
DICP : the annals of pharmacotherapy, 23(6), 445-450 (1989-06-01)
Loratadine is an addition to the class of nonsedating antihistamines which includes terfenadine, astemizole, and acrivastine. Loratadine is chemically related to the tricyclic antidepressants. Animal data have shown that insignificant amounts of loratadine enter the brain, and it has a
Safety and efficacy of loratadine in urticaria.
E W Monroe
International journal of dermatology, 35(12), 837-841 (1996-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service