Skip to Content
Merck
All Photos(1)

Key Documents

PHR1283

Supelco

Cefaclor

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Cefaclor monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14ClN3O4S · H2O
CAS Number:
Molecular Weight:
385.82
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 792
traceable to Ph. Eur. C0630000
traceable to USP 1096906

API family

cefaclor

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S1[C@H]2N(C(=C(C1)Cl)C(=O)O)C(=O)[C@H]2NC(=O)[C@H](N)c3ccccc3.O

InChI

1S/C15H14ClN3O4S.H2O/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7;/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23);1H2/t9-,10-,14-;/m1./s1

InChI key

WKJGTOYAEQDNIA-IOOZKYRYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Cefaclor belongs to the class of second generation semi-synthetic cephalosporin antibiotic drugs that is normally administered orally. With its broad-spectrum antibacterial action, cefaclor is effective against several gram-positive as well as gram-negative bacteria.

Application

Cefaclor may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and biological samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3323 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of cefaclor in human plasma by a sensitive and specific liquid chromatographic-tandem mass spectrometric method
Chen X, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 784(1), 17-24 (2003)
The use of a monolithic column to improve the simultaneous determination of four cephalosporin antibiotics in pharmaceuticals and body fluids by HPLC after solid phase extraction-a comparison with a conventional reversed-phase silica-based column
Samanidou VF, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 809(1), 175-182 (2004)
Determination of process-related impurities and degradation products in cefaclor by high-performance liquid chromatography
Lorenz LJ, et al.
Journal of Chromatographic Science, 30(6), 211-216 (1992)
Rapid and sensitive high-performance liquid chromatographic determination of four cephalosporin antibiotics in pharmaceuticals and body fluids
Samanidou VF, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 788(1), 147-158 (2003)
Simultaneous determination of cefpirome, cefaclor, ceftazidime, and cephradine in pharmaceutical formulations by reversed phase HPLC
Nawaz M, et al.
Acta Chromatographica , 23(2), 205-213 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service