Skip to Content
Merck
All Photos(1)

Key Documents

PHR1165

Supelco

Naproxen sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NaO3
CAS Number:
Molecular Weight:
252.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1457403

API family

naproxen

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].COc1ccc2cc(ccc2c1)[C@H](C)C([O-])=O

InChI

1S/C14H14O3.Na/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10;/h3-9H,1-2H3,(H,15,16);/q;+1/p-1/t9-;/m0./s1

InChI key

CDBRNDSHEYLDJV-FVGYRXGTSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Naproxen sodium (NS) is a sodium salt of propionic acid derivative and demonstrates analgesic, anti-pyretic, and anti-inflammatory properties. It exhibits efficiency in being used as an indomethacin in the treatment of acute musculoskeletal disorders. It belongs to the group of nonsteroidal anti-inflammatory drugs (NSAIDS) and acts by inhibiting the activity of both the cyclooxygenase enzymes, COX-1 & COX-2 and further blocking the synthesis of certain prostaglandins. It is administered in the treatment of ankylosing spondylitis, osteoarthritis, rheumatoid disorders, acute gout, mild to moderate pain, tendonitis, bursitis, dysmenorrhea, fever, and migraine headache. Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Naproxen sodium may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and combined dosage forms by chromatographic and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Cyclooxygenase (Prostaglandin H synthase 1 and 2) inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7178 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous Estimation of Sumatrirtan succinate and Naproxen Sodium in bulk drug and Pharmaceutical dosage form by RP-HPLC Method
Gondalia R and Dharamsi A
Journal of Drug Delivery and Therapeutics, 3(2), 1-6 (2013)
Simultaneous determination of naproxen sodium and acetaminophen in fixed-dose combinations formulations by first-order derivative spectroscopy: application to dissolution studies
Medina JR, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 7(3), 183-188 (2015)
Rapid simultaneous determination of sumatriptan succinate and naproxen sodium in combined tablets by validated ultra performance liquid chromatographic method
Reddy YR, et al.
Journal of Analytical and Bioanalytical Techniques , 2(3), 1-6 (2011)
Effects of preoperative ibuprofen and naproxen sodium on orthodontic pain
Polat O, et al.
The Angle Orthodontist, 75(5), 791-796 (2005)
Naproxen sodium, diflunisal, and placebo in the treatment of chronic back pain.
Berry H, et al.
Annals of the Rheumatic Diseases, 41(2), 129-132 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service