Skip to Content
Merck
All Photos(1)

Key Documents

PHR1153

Supelco

Triethyl citrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
Beilstein:
1801199
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1683606

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

API family

triethyl citrate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethyl citrate is an ester of citric acid and is commonly used as a plasticizer in pharmaceutical coatings including tablets, capsules, beads and granules for masking taste. It is also used as a flavoring agent in food.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Triethyl citrate can be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8988 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylcellulose--A Pharmaceutical Excipient with Multidirectional Application in Drug Dosage Forms Development
Wasilewska K and Winnicka K
Materials, 12(20), 3386-3386 (2019)
Oral Strip Technology: A review
Jaiswal, Hema
Indian Journal of Pharmaceutical and Biological Research, 2(2), 130-130 (2014)
Detection and quantification of low-molecular-weight aldehydes in pharmaceutical excipients by headspace gas chromatography
Li Z, et al.
Journal of Chromatography A, 1104(1-2), 1-10 (2006)
Development and validation of a gradient HPLC method for the determination of clindamycin and related compounds in a novel tablet formulation.
Platzer DJ, White BA.
Journal of Pharmaceutical and Biomedical Analysis, 41(1), 84-88 (2006)
M R Abbaspour et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 68(3), 747-759 (2007-11-06)
One challenge in tableting of sustained-release multiparticulates is maintaining the desired drug release after compaction. The aim of this study was to design sustained-release ibuprofen tablets which upon oral ingestion rapidly disintegrate into sustained-release pellets in which the integrity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service