Skip to Content
Merck
All Photos(1)

Key Documents

PHR1132

Supelco

Metoclopramide Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Metoclopramide HCl

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 357
traceable to Ph. Eur. M1825000
traceable to USP 1440808

API family

metoclopramide

CofA

current certificate can be downloaded

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Metoclopramide Hydrochloride is an antiemetic drug and is generally used for the treatment of gastrointestinal disorders. It can also exhibit sedative, extrapyrimidal, and prolactin secretion stimulation actions.

Application

Metoclopramide Hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and biological fluids by chromatography and chemiluminescence techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7152 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of stability indicating HPLC method for estimation of metoclopramide hydrochloride from a novel formulation
Shidhaye S, et al.
Journal of Pharmacy Research, 2(2), 290-295 (2009)
Flow-injection chemiluminescent determination of metoclopramide hydrochloride in pharmaceutical formulations and biological fluids using the [Ru (dipy) 32+]-permanganate system
Al-Arfaj NA
Talanta, 62(2), 255-263 (2004)
Rapid liquid chromatographic analysis of metoclopramide in pharmaceutical preparations
Aboul-Enein HY and Islam MR
Toxicological & Environmental Chemistry, 26(1-4), 197-201 (1990)
Assessment of a controlled release hydrophilic matrix formulation for metoclopramide HCl
Hasan EI, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 55(3), 339-344 (2003)
Formulation and evaluation of bi-layer tablet of metoclopramide hydrochloride and ibuprofen
Shiyani B, et al.
Aaps Pharmscitech, 9(3), 818-827 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service