Skip to Content
Merck
All Photos(1)

Key Documents

P3700000

Propylthiouracil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-Propyl-2-thiouracil, 2,3-Dihydro-6-propyl-2-thioxo-4(1H)-pyrimidinone, 4-Hydroxy-2-mercapto-6-propylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2OS
CAS Number:
Molecular Weight:
170.23
Beilstein:
130039
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

description

enzyme inhibitor

API family

propylthiouracil

manufacturer/tradename

EDQM

mp

218-220 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCC1=CC(=O)NC(=S)N1

InChI

1S/C7H10N2OS/c1-2-3-5-4-6(10)9-7(11)8-5/h4H,2-3H2,1H3,(H2,8,9,10,11)

InChI key

KNAHARQHSZJURB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Propylthiouracil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Annachiara Cavazzana et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 276(3), 753-759 (2019-01-04)
In clinical practice, when ability to perceive bitter taste is studied, quinine is preferred to phenylthiocarbamide (PTC) and 6-n-propylthiouracil (PROP) as taste stimulus, because many subjects are genetically non-tasters for PTC/PROP. However, it is poorly known how sensitive anterior (front)
A Rambaldi et al.
The Cochrane database of systematic reviews, (4)(4), CD002800-CD002800 (2005-10-20)
Randomised clinical trials have addressed the question whether propylthiouracil has any beneficial effects in patients with alcoholic liver disease. To assess the beneficial and harmful of propylthiouracil for patients with alcoholic liver disease. The Cochrane Hepato-Biliary Group Controlled Trials Register
C Vanek et al.
Thyroid : official journal of the American Thyroid Association, 15(1), 80-84 (2005-02-03)
Antineutrophil cytoplasmic antibodies (ANCA) are associated with vasculitis, including vasculitis induced by drugs such as the thionamides. The affected organ systems in thionamide-induced vasculitis have been primarily renal, musculoskeletal, and dermatologic. We describe the first case of thionamide-induced central nervous
Eleni Memi et al.
Journal of pediatric endocrinology & metabolism : JPEM, 25(3-4), 331-343 (2012-07-10)
Antithyroid drugs (ATDs) have been widely and effectively used for the treatment of pediatric and adult thyrotoxicosis for more than a half century. Since the very beginning of ATD use, reports of hepatic dysfunction related to propylthiouracil (PTU) therapy have
A Rambaldi et al.
Liver, 21(6), 398-404 (2002-03-21)
The aim of this review was to determine the benefits and adverse effects of propylthiouracil for patients with alcoholic liver disease. Systematic Cochrane Review of randomised clinical trials. The Cochrane Hepato-Biliary Controlled Clinical Trials Register, The Cochrane Library, MEDLINE, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service