P2939025
Prilocaine impurity E
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(RS)-N-(Methylphenyl)-2-(propylamino)propanamide
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
prilocaine
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
N(C(C)C(=O)Nc1ccc(cc1)C)CCC
InChI
1S/C13H20N2O/c1-4-9-14-11(3)13(16)15-12-7-5-10(2)6-8-12/h5-8,11,14H,4,9H2,1-3H3,(H,15,16)
InChI key
AQVLBLGVVXVVHN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service