Skip to Content
Merck
All Photos(1)

Key Documents

P2900000

Prednisone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Cortisone, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Dehydrocortisone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Beilstein:
2065301
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisone

manufacturer/tradename

EDQM

mp

236-238 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]([C@@](CC[C@@]3(C(CO)=O)O)([H])[C@]3(C)C4)([H])[C@]2([H])C4=O)=C1

InChI

1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1

InChI key

XOFYZVNMUHMLCC-ZPOLXVRWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednisone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles J Ryan et al.
The Lancet. Oncology, 16(2), 152-160 (2015-01-21)
Abiraterone acetate plus prednisone significantly improved radiographic progression-free survival compared with placebo plus prednisone in men with chemotherapy-naive castration-resistant prostate cancer at the interim analyses of the COU-AA-302 trial. Here, we present the prespecified final analysis of the trial, assessing
Claudine Angela Blum et al.
Lancet (London, England), 385(9977), 1511-1518 (2015-01-23)
Clinical trials yielded conflicting data about the benefit of adding systemic corticosteroids for treatment of community-acquired pneumonia. We assessed whether short-term corticosteroid treatment reduces time to clinical stability in patients admitted to hospital for community-acquired pneumonia. In this double-blind, multicentre
Fred Saad et al.
The Lancet. Oncology, 16(3), 338-348 (2015-02-24)
Orteronel is an investigational, partially selective inhibitor of CYP 17,20-lyase in the androgen signalling pathway, a validated therapeutic target for metastatic castration-resistant prostate cancer. We assessed orteronel in chemotherapy-naive patients with metastatic castration-resistant prostate cancer. In this phase 3, double-blind
Lindsay Sonstein et al.
The American journal of medicine, 127(11), 1097-1104 (2014-06-15)
Clinical practice guidelines recommend 40-60 mg of prednisone equivalent for 10-14 days for patients with acute exacerbations of chronic obstructive pulmonary disease (COPD). However, the amount of corticosteroid prescribed varies widely in clinical practice. Using the electronic health record, we
Katalin A Wilkinson et al.
Journal of immunology (Baltimore, Md. : 1950), 194(4), 1748-1754 (2015-01-16)
Tuberculosis-associated immune reconstitution inflammatory syndrome (TB-IRIS) frequently complicates combined antiretroviral therapy and antituberculosis therapy in HIV-1-coinfected tuberculosis patients. The immunopathological mechanisms underlying TB-IRIS are incompletely defined, and improved understanding is required to derive new treatments and to reduce associated morbidity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service