Skip to Content
Merck
All Photos(1)

Key Documents

NIST3673

Organic contaminants in non-smokers′ urine (frozen)

NIST® SRM® 3673

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

application(s)

clinical research

format

matrix material

storage temp.

−70°C

General description

Each unit of SRM 3673 consists of five vials each containing 10 mL of frozen urine.

SRM 3673_cert

SRM 3673_SDS

Application

Organic contaminants in non-smokers′ urine standard reference material (SRM) is intended for use in evaluating analytical methods used to determine selected hydroxylated polycyclic aromatic hydrocarbons (hydroxylated PAHs) and phthalate, phenol, and volatile organic compound (VOC) metabolites in urine.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction value of hydroxylated PAHs.
  • The non-certified values are included for phthalate, phenol, VOC metabolites, and organic analytes. 
  • Certified values are metrologically traceable to the International System of Units (SI) and SI-derived units of mass fraction. 
  • The NIST certificate is provided with storage, use and safety instructions.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service