Skip to Content
Merck
All Photos(1)

Key Documents

NIST2134

Arsenic implant in silicon depth profile standard

NIST SRM 2134

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of each

manufacturer/tradename

NIST®

application(s)

semiconductor

format

matrix material

General description

Each unit of SRM 2134 contains a single crystal silicon substrate, measuring 1 cm by 1 cm, which is ion-implanted with the isotope 75 As at an approximate energy of 100 keV.

SRM 2134_cert

SRM 2134_SDS

Application

The SRM is used to calibrate the secondary ion response to detect the minor and trace levels of arsenic within a silicon matrix using the analytical technique of secondary ion mass spectrometry (SIMS).

Features and Benefits

  • Available with a certificate documenting NIST-certified value for retained dose of 75As atoms and the dose is expressed in units of arsenic mass per unit area.
  • The reference and information values of elements are reported as mass fraction.
  • Certified values are retained dose of 75As atoms, and the dose is expressed in units of arsenic mass per unit area.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Arsenic (75As)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service