Skip to Content
Merck
All Photos(3)

Documents

M5921

Sigma-Aldrich

Magnesium sulfate heptahydrate

BioXtra, ≥99.0%

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38030409
PubChem Substance ID:
NACRES:
NA.25

vapor density

<0.01 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

product line

BioXtra

Assay

≥99.0%

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

solubility

H2O: 1 M, clear, colorless

cation traces

Al: ≤0.0005%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mn: ≤0.0005%
NH4+: ≤0.002%
Na: ≤0.005%
Pb: ≤0.0005%
Sr: ≤0.005%
Zn: ≤0.0005%

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium sulfate heptahydrate is hydroscopic and is vulnerable to warm temperatures.

Application

Magnesium sulfate heptahydrate has been used:
  • as a component in Kreb′s Ringer buffer
  • as a component in minimal medium to culture Ralstonia solanacearum
  • in the preparation of maceration solution for Lamb′s lettuce cell isolation and isotonic glucose buffer

Biochem/physiol Actions

Magnesium sulfate heptahydrate is used as a source of magnesium in foliar application of fertilizers to plants.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3D printing of plant tissue for innovative food manufacturing: Encapsulation of alive plant cells into pectin based bio-ink
Vancauwenberghe V, et al.
Journal of Food Engineering, 1-11 (2017)
Magnesium at Home: 25 Most Common Health Conditions & How Magnesium Salts Can Help, 27-27 (2018)
Comprehensive analysis of mitochondrial permeability transition pore activity in living cells using fluorescence-imaging-based techniques
Bonora M, et al.
Nature Protocols, 11(6), 1067-1067 (2016)
Exopolysaccharide Quantification for the Plant Pathogen Ralstonia solanacearum
Peyraud R, et al.
Bio-protocol, 7(10), 1067-1067 (2017)
Plant-Based Remediation Processes, 85-85 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service