Skip to Content
Merck
All Photos(1)

Documents

M2900000

Mometasone furoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(11β,16α)-9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30Cl2O6
CAS Number:
Molecular Weight:
521.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mometasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5ccco5)C(=O)CCl

InChI

1S/C27H30Cl2O6/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,22(32)14-28)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1

InChI key

WOFMFGQZHJDGCX-ZULDAHANSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mometasone furoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mometasone furoate is an anti-inflammatory glucocorticoid.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Svensson et al.
Primary care respiratory journal : journal of the General Practice Airways Group, 21(4), 412-418 (2012-09-25)
Acute rhinosinusitis is a common disease with an increasing incidence rate. It causes substantial costs to the individual and to society through healthcare consumption and absence from work. The use of antibiotics is widespread in the treatment of acute rhinosinusitis
S-H Wu et al.
The British journal of dermatology, 168(1), 172-178 (2012-07-28)
Lipoxins are potential anti-inflammatory mediators and serve as an endogenous 'braking signal' in the inflammatory process. Accumulating evidence has indicated the efficacy of lipoxin A(4) (LXA(4) ) and its analogs in the treatment of many animal models of inflammatory diseases.
David W Kennedy
Expert review of respiratory medicine, 6(5), 493-498 (2012-11-09)
Chronic rhinosinusitis is widely recognized as one of the most common chronic disease entities. Since its introduction in the USA in 1985, the role of functional endoscopic sinus surgery as an adjunct to medical therapy in the treatment of chronic
Environmental exposure unit: a sensitive, specific, and reproducible methodology for allergen challenge.
Anne K Ellis et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 111(5), 323-328 (2013-10-16)
Mohamed A Bitar et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(3), 931-937 (2012-09-27)
This study aimed at observing the efficacy of mometasone fuorate monohydrate nasal spray on obstructive adenoids in children and identifying the characteristics of responders using a pilot study including children aged 2-11 years, with evidence of more than 50 %

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service