Skip to Content
Merck
All Photos(1)

Key Documents

M1755200

Methylprednisolone hydrogen succinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methylprednisolone hemisuccinate, 11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-succinate, Methylprednisolone hydrogen succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34O8
CAS Number:
Molecular Weight:
474.54
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methylprednisolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)CCC(=O)OCC(=O)[C@@]1([C@@]2([C@H]([C@H]3[C@@H]([C@@]4(C(=CC(=O)C=C4)[C@H](C3)C)C)[C@H](C2)O)CC1)C)O

InChI

1S/C26H34O8.Na/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24;/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31);/q;+1/p-1/t14-,16-,17-,19-,23+,24-,25-,26-;/m0./s1

InChI key

FQISKWAFAHGMGT-SGJOWKDISA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylprednisolone hydrogen succinate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christos Krogias et al.
JAMA neurology, 70(8), 1056-1059 (2013-06-20)
A patient with a Caspr2 autoantibodies-associated syndrome had an unusual clinical triad and an excellent response to B-cell-anergizing therapy using the humanized monoclonal antibody tocilizumab directed against the interleukin 6 (IL-6) receptor. A 55-year-old man had an atypical clinical triad
Hirofumi Oyama et al.
Neurologia medico-chirurgica, 52(10), 769-773 (2012-10-26)
A 51-year-old female with a history of rheumatoid arthritis rapidly developed anterior neck pain and paresis in the left upper and lower extremities and right lower extremity, sensory disturbance in the left upper and lower extremities, and bladder and rectal
S Pescina et al.
International journal of pharmaceutics, 451(1-2), 12-17 (2013-05-01)
Trans-scleral iontophoresis, i.e. the application of small electric current to enhance drug transport across sclera is an option for non-invasive delivery of corticosteroids to the posterior segment of the eye. In this paper, in vitro trans-scleral iontophoresis of methylprednisolone hemisuccinate
Claudia Druschel et al.
Spine, 38(11), E669-E677 (2013-03-01)
Written mail-out survey. To determine current practice in high-dose methylprednisolone succinate (MPSS) administration for treatment of acute spinal cord injury (SCI) in Germany. Reanalysis of the National Acute Spinal Cord Injury Studies (NASCIS) resulted in criticism of the use of
Hoda Derakhshanian et al.
Canadian journal of physiology and pharmacology, 91(5), 380-385 (2013-05-10)
Glucocorticoid-induced osteoporosis (GIO) is the most common type of secondary osteoporosis. The aim of this study was to compare the efficacy of quercetin, a plant-derived flavonoid, with alendronate in the prevention of GIO. Fifty-six Sprague-Dawley rats were randomly distributed among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service