Skip to Content
Merck
All Photos(1)

Key Documents

M0178

Millipore

Middlebrook 7H9 Broth Base

suitable for microbiology, NutriSelect® Plus,

Recommended for use in isolation and cultivation of Mycobacterium species

Synonym(s):

Mycobacterial culture media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

final pH

6.6±0.2 (25 °C)

application(s)

clinical testing
food and beverages
life science and biopharma
veterinary

microbiology

storage temp.

2-25°C

suitability

mycobacteria

General description

Middlebrook 7H9 Broth Base contains inorganic compounds that supply the elements potassium, sulfur, magnesium, and phosphorus which are required for appropriate growth of Mycobacteria. This medium with Middlebrook ADC Growth Supplement is recommended for ideal growth of Mycobacterium species.
Middlebrook ADC Growth Supplement (Product No. M0553) contains bovine albumin, dextrose, and catalase. Dextrose supplies energy, while albumin protects Mycobacteria from toxic agents and catalase splits toxic radicals.
Mycobacteria grow most rapidly in broth media and hence primary isolation of bacterial species can be carried out in Middlebrook 7H9 Broth Base.

Application

Middlebrook 7H9 Broth Base has been used for the cultivation Mycobacterium bovis bacillus Calmette-Guérin Pasteur strain. It is also recommended for cultivation and sensitivity testing of Mycobacterium tuberculosis.

Components

Ingredients (g/L)
Ammonium sulfate, 0.50
Disodium phosphate, 2.50
Monopotassium phosphate, 1.00
Sodium citrate, 0.10
Magnesium sulfate, 0.05
Calcium chloride, 0.0005
Zinc sulfate, 0.001
Copper sulfate, 0.001
Ferric ammonium citrate, 0.04
L-Glutamic acid, 0.50
Pyridoxine, 0.001
Biotin, 0.0005

Preparation Note

Suspend 2.35 g of Middlebrook 7H9 Broth Base in 450 ml of distilled water. Add either 2 ml of glycerol (Cat. No 49767) or 0.5 g of TWEEN® 80 (Cat. No. 93780). Heat if necessary to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. Pressure (121°C) for 10 minutes. Cool to 45°C or below and aseptically add 1 vial of Middlebrook ADC Growth Supplement (Cat. No. M0553). Mix well before dispensing.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Masoko et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 586247-586247 (2013-08-21)
The aim of the study was to scientifically evaluate the antimycobacterial activity of selected indigenous medicinal plants from the Limpopo Province used for the treatment of humans with symptoms of Mycobacterium tuberculosis. The leaves of five plant species (Apodytes dimidiata
Differential requirements for L-citrulline and L-arginine during antimycobacterial macrophage activity
Rapovy SM, et al.
Journal of Immunology, 195(7), 3293-3300 (2015)
Nancy Patience Motlalepula Komape et al.
BMC complementary and alternative medicine, 17(1), 128-128 (2017-02-27)
Tuberculosis is an infectious communicable disease and the causative agent of the disease has over the years developed resistance to streamline chemotherapeutic agents with dire consequences and there is a need for development of new and more potent alternatives. Constituents
l-Citrulline Metabolism in Mice Augments CD4+ T Cell Proliferation and Cytokine Production In Vitro, and Accumulation in the Mycobacteria-Infected Lung
Lange S M, et al.
Frontiers in Immunology, 8, 1561-1561 (2017)
Daniel J Greenwood et al.
Science (New York, N.Y.), 364(6447), 1279-1282 (2019-06-30)
Tuberculosis, caused by the intracellular pathogen Mycobacterium tuberculosis, remains the world's deadliest infectious disease. Sterilizing chemotherapy requires at least 6 months of multidrug therapy. Difficulty visualizing the subcellular localization of antibiotics in infected host cells means that it is unclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service