Skip to Content
Merck
All Photos(3)

Key Documents

K2012

Sigma-Aldrich

Ketoprofen

meets USP testing specifications

Synonym(s):

2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

CC(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)

InChI key

DKYWVDODHFEZIM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application


  • A strong, silk protein-inspired tissue adhesive with an enhanced drug release mechanism for transdermal drug delivery.: Although not exclusively focused on ketoprofen, this study details a novel adhesive for transdermal delivery, highlighting methods that could be adapted for enhancing the delivery of ketoprofen, thus being of interest to biochemists specializing in transdermal drug formulations (Fang et al., 2024).

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is selective for COX-1.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Christiansen et al.
Journal of medicinal chemistry, 54(19), 6691-6703 (2011-08-23)
The free fatty acid receptor 1 (FFA1, also known as GPR40) enhances glucose-stimulated insulin secretion from pancreatic β-cells and is recognized as an interesting new target for treatment of type 2 diabetes. Several series of selective FFA1 agonists are already
Nathalie C Newby et al.
Journal of dairy science, 96(3), 1511-1520 (2013-01-22)
Surgical correction of left displaced abomasum (LDA) is common in lactating dairy cattle. Despite the growing acceptance that abdominal surgery is painful, few cows are administered analgesia following LDA surgery. The objective of this research was to examine the effect
Ikram Ullah Khan et al.
International journal of pharmaceutics, 441(1-2), 809-817 (2012-12-26)
Microchannels based microfluidic systems are able to obtain monodispersed microparticles but are limited by cost, time and channel clogging. We succeeded in on the fly encapsulation of high ketoprofen contents in acrylate-based copolymer microbeads by environment friendly UV induced free
M Sugiura et al.
Contact dermatitis, 43(1), 16-19 (2000-07-21)
We report 4 cases of photocontact dermatitis due to ketoprofen, a non-steroidal anti-inflammatory drug derived from propionic acid. We carried out a skin biopsy to examine the amount of ketoprofen in the eruptive skin. We investigated the cross-reactions between tiaprofenic
F Jamali et al.
Clinical pharmacokinetics, 19(3), 197-217 (1990-09-01)
Ketoprofen, a potent nonsteroidal anti-inflammatory drug (NSAID) of the 2-arylpropionic acid class, has been used clinically for over 15 years in Europe, and has recently been introduced in the United States. Although it possesses a chiral centre, with only the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service