Skip to Content
Merck
All Photos(1)

Key Documents

IRMM643

Sulfur-32S solution

IRMM®, certified reference material, Spike Isotopic Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
32S
CAS Number:
Molecular Weight:
31.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material

Agency

IRMM®

isotopic purity

 32S

manufacturer/tradename

JRC

concentration

371.96 μmol/kg±0.57 μmol/kg 32S in 2.8 M HNO3

application(s)

general analytical

format

single component solution

mass shift

depleted

SMILES string

[32S]

InChI

1S/S/i1+0

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM643

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Chen et al.
PloS one, 9(9), e105400-e105400 (2014-09-03)
Hydrogen sulfide (H2S), as a potential gaseous messenger molecule, has been suggested to play important roles in a wide range of physiological processes in plants. The aim of present study was to investigate which set of proteins is involved in
Sudhanshu S Pawar et al.
Bioresource technology, 169, 677-685 (2014-08-12)
Caldicellulosiruptor saccharolyticus has gained reputation as being among the best microorganisms to produce H2 due to possession of various appropriate features. The quest to develop an inexpensive cultivation medium led to determine a possible replacement of the expensive component cysteine
Beata Olas et al.
Chemico-biological interactions, 220, 20-24 (2014-06-15)
Hydrogen sulfide (H2S) is a well known toxic gas at high levels. However, at physiological levels, H2S may play a role in the pathogenesis of various cardiovascular diseases. The objective was to study the effects of exogenous H2S on the
Yusuke Kumai et al.
Pflugers Archiv : European journal of physiology, 467(4), 651-664 (2014-06-19)
The present study investigated the role of hydrogen sulfide (H2S) in regulating Na(+) uptake in larval zebrafish, Danio rerio. Waterborne treatment of larvae at 4 days post-fertilization (dpf) with Na2S or GYY-4137 (chemicals known to generate H2S) significantly reduced Na(+)
Dae-Hwan Kim et al.
Nanoscale, 6(23), 14549-14554 (2014-10-29)
A high-quality Sb₂S₃ thin-absorber with controllable thickness was reproducibly formed by atomic layer deposition (ALD) technique. Compared with conventional chemical bath deposition (CBD), the Sb₂S₃ absorber deposited by ALD did not contain oxide or oxygen impurities and showed a very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service