Skip to Content
Merck
All Photos(1)

Documents

I7000000

Itraconazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Oriconazole, R51211, Sporanox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H38Cl2N8O4
CAS Number:
Molecular Weight:
705.63
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

itraconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(C)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OCC5COC(Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4

InChI

1S/C35H38Cl2N8O4/c1-3-25(2)45-34(46)44(24-40-45)29-7-5-27(6-8-29)41-14-16-42(17-15-41)28-9-11-30(12-10-28)47-19-31-20-48-35(49-31,21-43-23-38-22-39-43)32-13-4-26(36)18-33(32)37/h4-13,18,22-25,31H,3,14-17,19-21H2,1-2H3

InChI key

VHVPQPYKVGDNFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Itraconazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Itraconazole inhibits cytochrome P-450-dependent enzymes which results in the inhibition of ergosterol synthesis. It does so by interacting with 14-α demethylase, which is a cytochrome P-450 enzyme necessary to convert lanosterol to ergosterol. Ergosterol is a crucial compenent of fungal cell membranes. Therefore, it′s inhibition results in increased cellular permeability causing leakage of cellular contents. Itraconazole may also inhibit endogenous respiration, interact with membrane phospholipids, inhibit the transformation of yeasts to mycelial forms, inhibit purine uptake, and impair triglyceride and phospholipid biosynthesis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zsombor K Nagy et al.
International journal of pharmaceutics, 480(1-2), 137-142 (2015-01-18)
High speed electrospinning (HSES), compatible with pharmaceutical industry, was used to demonstrate the viability of the preparation of drug-loaded polymer nanofibers with radically higher productivity than the known single-needle electrospinning (SNES) setup. Poorly water-soluble itraconazole (ITRA) was formulated with PVPVA64
A O S Fonseca et al.
The Journal of antimicrobial chemotherapy, 69(6), 1564-1567 (2014-02-14)
The purpose of this study was to compare the in vitro susceptibilities of 22 Brazilian isolates of Pythium insidiosum to antifungals using a standardized inoculum of zoospores and a proposed novel inoculum prepared from cultured mycelia (hyphae) of P. insidiosum.
Ritesh Agarwal et al.
Mycoses, 56(5), 559-570 (2013-03-19)
Patients with aspergilloma can be safely managed with supportive therapy in absence of massive haemoptysis. We hypothesised that chronic cavitary pulmonary aspergillosis (CCPA) could also be managed on similar grounds. The aim of this prospective, randomised controlled trial was to
James D Homans et al.
The Pediatric infectious disease journal, 29(1), 65-67 (2009-11-04)
Coccidioides immitis causes a wide range of disease in humans. Fluconazole and itraconazole are effective treatments. Clinical evidence suggests that itraconazole is equivalent or superior to fluconazole in treating osteoarticular infections in rates of cure and recurrence. We report 2
Ji-Qin Wu et al.
Antimicrobial agents and chemotherapy, 58(8), 4464-4469 (2014-05-29)
Amphotericin B (AMB) has been a mainstay therapy for fungal infections of the central nervous system, but its use has been limited by its poor penetration into the brain, the mechanism of which remains unclear. In this study, we aimed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service