Skip to Content
Merck
All Photos(1)

Documents

H1450000

Hyoscine butylbromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Scopolamine N-butyl bromide, (−)-N-Butylscopolamine bromide, Hyoscine N-butyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30BrNO4
CAS Number:
Molecular Weight:
440.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hyoscine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].[H][C@@]12C[C@H](C[C@@]([H])([C@]3([H])O[C@]13[H])[N+]2(C)CCCC)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C21H30NO4.BrH/c1-3-4-10-22(2)17-11-15(12-18(22)20-19(17)26-20)25-21(24)16(13-23)14-8-6-5-7-9-14;/h5-9,15-20,23H,3-4,10-13H2,1-2H3;1H/q+1;/p-1/t15-,16-,17-,18+,19-,20+,22?;/m1./s1

InChI key

HOZOZZFCZRXYEK-GSWUYBTGSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hyoscine butylbromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timur H Kuru et al.
Journal of computer assisted tomography, 38(4), 558-564 (2014-04-16)
To compare 2 previously presented algorithms for extracting parameters from intravoxel incoherent motion (IVIM) studies and investigate them in the context of tissue differentiation. Magnetic resonance imaging (MRI) was performed in 23 patients without histologically proven prostate carcinoma (PCa) and
Guido N Tytgat
Current medical research and opinion, 24(11), 3159-3173 (2008-10-15)
Being a quaternary ammonium compound derived from scopolamine, the alkaloid hyoscine butylbromide (HBB) exerts anticholinergic effects without side effects related to the central nervous system because it does not pass the blood-brain barrier. Clinical experience with this antispasmodic dates back
Guido N Tytgat
Drugs, 67(9), 1343-1357 (2007-06-06)
Abdominal cramping and pain is a frequent problem in the adult population of Western countries, with an estimated prevalence of < or =30%. Hyoscine butylbromide (scopolamine butylbromide) [Buscopan/Buscapina] is an antispasmodic drug indicated for the treatment of abdominal pain associated
Andrew A Plumb et al.
AJR. American journal of roentgenology, 203(4), W400-W407 (2014-09-24)
The purpose of this article is to describe the MRI findings associated with lymphoid nodular hyperplasia at MR enterography and test the ability of radiologists to differentiate healthy control subjects from patients with Crohn disease (CD). Ethical approval was granted
K Mackway-Jones et al.
Emergency medicine journal : EMJ, 20(3), 267-267 (2003-05-16)
A short cut review was carried out to establish whether buscopan (hyoscine butylbromide) is better than analgesics at controlling pain in abdominal colic. A total of 31 papers were found using the reported search, of which none presented any evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service