Skip to Content
Merck
All Photos(1)

Key Documents

G0700000

Guaifenesin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Guaiacol glyceryl ether, 3-(o-Methoxyphenoxy)-1,2-propanediol, Glycerol guaiacolate, Guaifenesin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O4
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

guaifenesin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccccc1OCC(O)CO

InChI

1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-7-8(12)6-11/h2-5,8,11-12H,6-7H2,1H3

InChI key

HSRJKNPTNIJEKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Guaifenesin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Robert R Fayzullin et al.
Journal of pharmaceutical sciences, 103(10), 3176-3182 (2014-08-06)
The solubility of 3-(2-methoxyphenoxy)-propane-1,2-diol, the well-known chiral drug guaifenesin 1, in water has been investigated by means of polythermal and isothermal approaches. It was found that the solubilities of racemic and enantiomeric diols rac- and (R)-1 depend strongly on temperature.
William Storms et al.
Current allergy and asthma reports, 9(2), 101-106 (2009-02-13)
Mucus in the airways is a complex mixture of water, lipids, glycoproteins, sugars, and electrolytes that serves as a lubricant for the epithelium. The efficient flow of respiratory mucus is a first level of immune defense that requires an appropriate
Manujendra Ray et al.
Hospital practice (1995), 37(1), 60-63 (2010-09-30)
Adverse drug reactions lead to a significant number of hospital admissions each year and thus contribute to the overall financial burden of health care. Some of these drug reactions are allergic responses. As the overall predictability of allergic responses to
Stephen Bennett et al.
Journal of alternative and complementary medicine (New York, N.Y.), 10(6), 967-969 (2005-01-28)
Ephedrine and guaifenesin are herbal supplements that have experienced increased use over the past decade. Ephedrine has been used as a stimulant and weight-loss product, guaifenesin as an expectorant and cough suppressant; both are found in combination in many antitussives
Clinical inquiries. Is guaifenesin safe during pregnancy?
Roberto Silva et al.
The Journal of family practice, 56(8), 669-670 (2007-08-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service