Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
API family
glipizide
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[S](=O)(=O)(N)c1ccc(cc1)CCNC(=O)c2ncc(nc2)C
InChI
1S/C14H16N4O3S/c1-10-8-18-13(9-17-10)14(19)16-7-6-11-2-4-12(5-3-11)22(15,20)21/h2-5,8-9H,6-7H2,1H3,(H,16,19)(H2,15,20,21)
InChI key
IMEZLHZLIANIAS-UHFFFAOYSA-N
General description
Application
Packaging
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename EDQM | manufacturer/tradename EDQM | manufacturer/tradename EDQM | manufacturer/tradename EDQM |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| API family glipizide | API family glipizide | API family glipizide | API family glipizide |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

