Skip to Content
Merck
All Photos(4)

Key Documents

F7878

Supelco

Folinic acid calcium salt hydrate

analytical standard

Synonym(s):

5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt, 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt, 5-HCO-H4PteGlu, Calcium folinate, Citrovorum factor calcium salt, Leucovorin calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21CaN7O7 · xH2O
CAS Number:
Molecular Weight:
511.50 (anhydrous basis)
Beilstein:
5723924
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic (organic)

Quality Level

grade

analytical standard

Assay

≥90.0%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

yellow to orange

mp

240-250 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

[Ca++].[H]O[H].[H]C(=O)N1C(CNc2ccc(cc2)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)CNc3nc(N)nc(O)c13

InChI

1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1

InChI key

KVUAALJSMIVURS-ZEDZUCNESA-L

Looking for similar products? Visit Product Comparison Guide

General description

Folinic acid calcium salt hydrate is a drug, widely used for the treatment of cancer, since it can reduce toxic side effects and helps to improve drug accumulation in tumor tissues.

Application

Calcium folinate offsets the effects of folic acid antagonists that act by binding dihydrofolate reductase and blocking the conversion of folic acid to tetrahydrofolate. In medicine, calcium folinate is used after treatment with methotrexate to reduce the toxicity of the methotrexate. Calcium folinate is also used to enhance the cytotoxic effect of 5-fluorouracil in the treatment of cancer.
Folinic acid calcium salt hydrate has been used as a test compound to study its effects on microglia involved in neuro-inflammation using enzyme-linked immunosorbent assay (ELISA) technique.
Folinic acid calcium salt hydrate has been used as a test compound to study its effects on microglia involved in neuro-inflammation using enzyme-linked immunosorbent assay (ELISA) technique. Folinic acid calcium salt hydrate may be used as an analytical reagent for the quantification of methotrexate its metabolites in biological samples using turbulent flow chromatography with tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The combination of vitamins and omega-3 fatty acids has an enhanced anti-inflammatory effect on microglia
Kurtys E, et al.
Neurochemistry International, 99, 206-214 (2016)
Preparation, characterization and physicochemical properties of DOPE-PEG2000 stabilized oleic acid-soy lecithin liposomes (POLL)
Suk ERV and Misran M
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 513, 267-273 (2017)
Development and validation of a turbulent flow chromatography and tandem mass spectrometry method for the quantitation of methotrexate and its metabolites 7-hydroxy methotrexate and DAMPA in serum
Schofield.CR, et al.
Journal of Chromatography. B, Biomedical Applications, 1002, 169-175 (2015)
William Greenhalf et al.
Journal of the National Cancer Institute, 106(1), djt347-djt347 (2013-12-05)
Human equilibrative nucleoside transporter 1 (hENT1) levels in pancreatic adenocarcinoma may predict survival in patients who receive adjuvant gemcitabine after resection. Microarrays from 434 patients randomized to chemotherapy in the ESPAC-3 trial (plus controls from ESPAC-1/3) were stained with the
Pasquale F Innominato et al.
Cancer, 119(14), 2564-2573 (2013-05-02)
Chemotherapy-induced neutropenia has been associated with prolonged survival selectively in patients on a conventional schedule (combined 5-fluorouracil, leucovorin, and oxaliplatin [FOLFOX2]) but not on a chronomodulated schedule of the same drugs administered at specific circadian times (chronoFLO4). The authors hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service