Skip to Content
Merck
All Photos(1)

Key Documents

E2470000

Etodolac

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, Etodolic acid, NSC 282126

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3
CAS Number:
Molecular Weight:
287.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

etodolac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCc1cccc2c3CCOC(CC)(CC(O)=O)c3[nH]c12

InChI

1S/C17H21NO3/c1-3-11-6-5-7-12-13-8-9-21-17(4-2,10-14(19)20)16(13)18-15(11)12/h5-7,18H,3-4,8-10H2,1-2H3,(H,19,20)

InChI key

NNYBQONXHNTVIJ-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etodolac EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is a non-selective inhibitor of COX-1 and COX-2.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etodolac: the chemistry, pharmacology, metabolic disposition, and clinical profile of a novel anti-inflammatory pyranocarboxylic acid.
L G Humber
Medicinal research reviews, 7(1), 1-28 (1987-01-01)
Vesile Öztürk et al.
Pain practice : the official journal of World Institute of Pain, 13(3), 191-197 (2012-06-27)
We aimed to determine the efficacy and safety of etodolac, in acute migraine attacks in comparison with paracetamol (acetaminophen). We designed a randomized, double-blind, crossover phase III clinical trial for patients diagnosed with migraine for at least 1 year, according
M Pena
Rheumatology international, 10 Suppl, 9-16 (1990-01-01)
Numerous clinical trials have shown etodolac to be an effective analgesic. The purpose of the present report is to review results of 14 studies that demonstrate the effectiveness of etodolac in a variety of painful conditions. Presented are the results
Z Adhim et al.
British journal of cancer, 105(3), 393-402 (2011-07-14)
Although the anti-tumour effect of cyclooxygenase-2 (Cox-2) inhibitors in invasive bladder cancer has been confirmed, its mechanisms of action are unclear. Recently, the concept of an epithelial-to-mesenchymal transition (EMT) promoting carcinoma progression has been suggested, and a key feature of
Soad S Abd el-Hay et al.
Journal of fluorescence, 22(1), 247-252 (2011-08-20)
A simple, selective and sensitive luminescence method has been developed for the assay of etodolac (I), moxepril HCl (II) and fexofenadine HCl (III) in bulk drug and pharmaceutical formulations. The method is based on the luminescence sensitization of europium (Eu(3+))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service