Skip to Content
Merck
All Photos(1)

Key Documents

E2028

Sigma-Aldrich

Ethidium bromide monoazide

≥95% purity (HPLC), solid

Synonym(s):

3-Amino-8-azido-5-ethyl-6-phenylphenanthridinium bromide, Ethidium monoazide bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18BrN5
CAS Number:
Molecular Weight:
420.31
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Ethidium bromide monoazide, ≥95% (HPLC), solid

Quality Level

Assay

≥95% (HPLC)

form

solid

technique(s)

titration: suitable

color

orange

solubility

DMF: soluble
ethanol: soluble

ε (extinction coefficient)

≥5200 at 459-465 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Br-].CC[n+]1c(-c2ccccc2)c3cc(ccc3c4ccc(N)cc14)N=[N+]=[N-]

InChI

1S/C21H17N5.BrH/c1-2-26-20-12-15(22)8-10-18(20)17-11-9-16(24-25-23)13-19(17)21(26)14-6-4-3-5-7-14;/h3-13,22H,2H2,1H3;1H

InChI key

GHUXAYLZEGLXDA-UHFFFAOYSA-N

Biochem/physiol Actions

Photoactive stain covalently binds to nucleic acids in solution and in cells with damaged membranes.
Used to footprint drug binding sites on DNA, detect non-viable cells, identify ethidium binding sites on DNA and tRNA, and selectively inactivate gene expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

C-W Chang et al.
Indoor air, 21(4), 291-299 (2011-01-05)
Legionella pneumophila, aerosolized from numerous indoor facilities (e.g., shower heads, hot tubs, spas), may cause Pontiac fever (PF) and lethal pneumonia named Legionnaires' disease (LD) in humans. Reliable methods on quantitative exposure assessment of this bioaerosol are essential for the
Sara Daniel et al.
Clinical epigenetics, 10(1), 138-138 (2018-11-08)
Cancer treatments have substantially improved childhood cancer survival but are accompanied by long-term complications, notably chronic inflammatory diseases. We hypothesize that cancer treatments could lead to long-term epigenetic changes in immune cells, resulting in increased prevalence of inflammatory diseases in
Ji-Min Ju et al.
Nature communications, 9(1), 225-225 (2018-01-18)
Whether hematopoietic cell-restricted distribution of antigens affects the degree of thymic negative selection has not been investigated in detail. Here, we show that T cells specific for hematopoietic cell-restricted antigens (HRA) are not completely deleted in the thymus, using the
G Loozen et al.
Molecular oral microbiology, 26(4), 253-261 (2011-07-07)
DNA-based methodology for the identification and detection of specific bacteria in dental plaque offers advantages over culturing techniques. One drawback of current molecular techniques like real-time quantitative polymerase chain reaction (RT-QPCR) is that they are not able to distinguish between
Mark Blooi et al.
Fungal biology, 117(2), 156-162 (2013-03-05)
Detection of the lethal amphibian fungus Batrachochytrium dendrobatidis relies on PCR-based techniques. Although highly accurate and sensitive, these methods fail to distinguish between viable and dead cells. In this study a novel approach combining the DNA intercalating dye ethidium monoazide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service