Skip to Content
Merck
All Photos(1)

Key Documents

E1400000

Erythromycin estolate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Erythromycin 2′-propionate dodecyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C40H71NO14 · C12H26SO4
CAS Number:
Molecular Weight:
1056.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

erythromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCOS(O)(=O)=O.CC[C@H]1OC(=O)[C@@H](C)[C@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@@H](C)[C@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(O)C[C@H](C)C(=O)C(C)[C@@H](O)[C@@]1(C)O

InChI

1S/C37H67NO13.C12H26O4S/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2-12H2,1H3,(H,13,14,15)/t18-,19+,20?,21+,22-,23-,24-,25?,26-,28+,29+,30+,31-,32-,34-,35-,36+,37+;/m0./s1

InChI key

SKDGGFHGLZBNBC-CAHOBLCESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Erythromycin estolate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Pari et al.
Pharmacological research, 49(5), 481-486 (2004-03-05)
Tetrahydrocurcumin (THC), one of the major metabolites of curcumin, was investigated for its possible hepatoprotective effect in Wistar rats against erythromycin estolate-induced toxicity. Oral administration of THC significantly prevented the occurrence of erythromycin estolate-induced liver damage. The increased level of
Soo-Jin Park et al.
Journal of colloid and interface science, 271(2), 336-341 (2004-02-20)
In this work, the producing of a biodegradable poly(l-lactide) (PLA)/poly(ethylene glycol) (PEG) microcapsule by emulsion solvent evaporation method was investigated. The effect of PEG segments added to the PLA microcapsules on the degradation, size distribution, and release behavior was studied.
Cholestatic hepatitis caused by esters of erythromycin and oleandomycin. 1962.
F I Gilbert
Hawaii medical journal, 54(6), 603-605 (1995-06-11)
Pidaran Murugan et al.
Journal of basic and clinical physiology and pharmacology, 16(1), 1-15 (2005-09-29)
Erythromycin estolate (EME), a potent macrolide antibiotic, generates free radicals, but their role in the development of liver toxicity is not yet well understood. The present study was carried out to investigate the effect of the antioxidant drug tetrahydrocurcumin (a
[Hepatotoxicity from erythromycin estolate].
S Saiz Santos et al.
Atencion primaria, 10(9), 1050-1050 (1992-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service