Skip to Content
Merck
All Photos(3)

Key Documents

D9564

Sigma-Aldrich

DAPI, dilactate

≥98% purity (HPLC), powder

Synonym(s):

4′,6-Diamidino-2-phenyindole, dilactate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N5 · 2C3H6O3
Molecular Weight:
457.48
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

DAPI, dilactate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

titration: suitable

color

yellow

solubility

H2O: 20 mg/mL

ε (extinction coefficient)

28.00-32.50 at 345-351 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CC(O)C(O)=O.CC(O)C(O)=O.NC(=N)c1ccc(cc1)-c2cc3ccc(cc3[nH]2)C(N)=N

InChI

InChI=1S/C16H15N5.C3H6O3/c17-15(18)10-3-1-9(2-4-10)13-7-11-5-6-12(16(19)20)8-14(11)21-13;1-2(4)3(5)6/h1-8,21H,(H3,17,18)(H3,19,20);2,4H,1H3,(H,5,6)

InChI key

LCAUPANIXBMNIA-UHFFFAOYSA-N

General description

DAPI is an indolenine dye. It carries two amidine components, one on a pendant phenyl ring and another directly on the indole moiety.

Application

DAPI, dilactate has been used for the nuclear staining in cells. DAPI is a common flow cytometry stain that is widely used as a DNA and nuclear stain. Additionally, it is used as a counterstain following in situ hybridization and other immunofluorescence procedures.

Biochem/physiol Actions

A high sensitivity dye used to detect single nucleic acid molecules. DAPI has been shown to detect a single DNA molecule in solution, and to detect femtograms of DNA in single cells and chloroplasts.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahrzad Bahrampour et al.
PLoS biology, 17(2), e3000163-e3000163 (2019-02-27)
During central nervous system (CNS) development, genetic programs establish neural stem cells and drive both stem and daughter cell proliferation. However, the prominent anterior expansion of the CNS implies anterior-posterior (A-P) modulation of these programs. In Drosophila, a set of
Expression and high glucose-mediated regulation of K+ channel interacting protein 3 (KChIP3) and KV4 channels in retinal Muller glial cells.
Chavira-Suarez E, et al.
Biochemical and Biophysical Research Communications, 404, 678-683 (2011)
Vicky Goler-Baron et al.
PloS one, 6(1), e16007-e16007 (2011-02-02)
Multidrug resistance (MDR) is a major impediment to curative cancer chemotherapy. The ATP-Binding Cassette transporters ABCG2, ABCB1 and ABCC2 form a unique defense network against multiple structurally and functionally distinct chemotherapeutics, thereby resulting in MDR. Thus, deciphering novel mechanisms of
Nicolas Buchon et al.
BMC biology, 8, 152-152 (2010-12-24)
Gut homeostasis is central to whole organism health, and its disruption is associated with a broad range of pathologies. Following damage, complex physiological events are required in the gut to maintain proper homeostasis. Previously, we demonstrated that ingestion of a
Silvia S Kang et al.
The Journal of experimental medicine, 208(4), 747-759 (2011-04-06)
After virus infection, cytotoxic T lymphocytes (CTLs) divide rapidly to eradicate the pathogen and prevent the establishment of persistence. The magnitude of an antiviral CTL response is thought to be controlled by the initiation of a cell cycle program within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service