Skip to Content
Merck
All Photos(2)

Key Documents

D9257

Supelco

Dihydrotachysterol

analytical standard

Synonym(s):

9,10-Secoergosta-5,7,22-trien-3β-ol, DHT2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]([C@@]1([H])CC[C@@]2([H])/C(CCC[C@@]21C)=C/C=C3[C@@H](C)CC[C@H](O)C/3)/C=C/[C@H](C)C(C)C

InChI

1S/C28H46O/c1-19(2)20(3)12-13-22(5)25-16-17-26-23(10-8-18-28(25,26)6)14-15-24-21(4)9-7-11-27(24)29/h12-15,19-22,25-27,29H,7-11,16-18H2,1-6H3/b13-12+,23-14+,24-15+/t20-,21-,22+,25?,26?,27-,28+/m0/s1

InChI key

DTSXXSAWQHPLEF-GFVAUXBKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrotachysterol is a vitamin D analog in the reduced form, which has implications in the studies of calcemic and cell differentiating action of the hormone, 1α,25-dihydroxyvitamin D3. It has been indicated as a medication for the treatment of hypoparathyroidism and renal osteodystrophy.

Application

Dihydrotachysterol may be used as an analytical reference standard for the determination of the analyte in biological samples by chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum dihydrotachysterol levels and biological action in normal man
TAYLOR A, et al.
The Journal of clinical endocrinology and metabolism, 67(1), 198-202 (1988)
Gas chromatographic studies of vitamins D2 and D3
Nair PP, et al.
Analytical Chemistry, 37(6), 631-636 (1965)
Ali Mohammad Tohari et al.
Experimental cell research, 392(1), 112023-112023 (2020-04-24)
Diabetic retinopathy (DR) is a diabetes-associated complication characterized by irreversible deterioration of the microvessels within the retina, leading subsequently to severe retinal damage and vision loss. Vitamin D (VITD), a steroid hormone, plays multiple physiological functions in cellular homeostasis. Deficiency
Signe Kjelstrup et al.
Biophysical journal, 96(11), 4376-4386 (2009-06-03)
Coefficients for active transport of ions and heat in vesicles with Ca(2+)-ATPase from sarcoplasmic reticulum are defined in terms of a newly proposed thermodynamic theory and calculated using experiments reported in the literature. The coefficients characterize in a quantitative manner
[Cerebral calcinosis].
J G Heckmann et al.
Medizinische Klinik (Munich, Germany : 1983), 96(1), 55-56 (2001-02-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service