Skip to Content
Merck
All Photos(1)

Documents

D0950000

Diazoxide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClN2O2S
CAS Number:
Molecular Weight:
230.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diazoxide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1=Nc2ccc(Cl)cc2S(=O)(=O)N1

InChI

1S/C8H7ClN2O2S/c1-5-10-7-3-2-6(9)4-8(7)14(12,13)11-5/h2-4H,1H3,(H,10,11)

InChI key

GDLBFKVLRPITMI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diazoxide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa M Anastacio et al.
Journal of the American College of Surgeons, 216(6), 1144-1149 (2013-03-29)
Diazoxide maintains myocyte volume and contractility during stress via an unknown mechanism. The mechanism of action may involve an undefined (genotype unknown) mitochondrial ATP-sensitive potassium channel and is dependent on the ATP-sensitive potassium channel subunit sulfonylurea type 1 receptor (SUR1).
Bernard Pirotte et al.
Journal of medicinal chemistry, 56(8), 3247-3256 (2013-03-23)
The synthesis of diversely substituted 3-alkyl/aralkyl/arylamino-1,4,2-benzodithiazine 1,1-dioxides and 3-alkylaminopyrido[4,3-e]-1,4,2-dithiazine 1,1-dioxides is described. Their biological activities on pancreatic β-cells and on smooth muscle cells were compared to those of the reference ATP-sensitive potassium channel (KATP channel) openers diazoxide and 7-chloro-3-isopropylamino-4H-1,2,4-benzothiadiazine 1,1-dioxide.
Ekhson L Holmuhamedov et al.
PloS one, 7(9), e44667-e44667 (2012-09-14)
Cardiac subsarcolemmal (SSM) and interfibrillar (IFM) mitochondrial subpopulations possess distinct biochemical properties and differ with respect to their protein and lipid compositions, capacities for respiration and protein synthesis, and sensitivity to metabolic challenge, yet their responsiveness to mitochondrially active cardioprotective
Yael Levy-Shraga et al.
Journal of pediatric endocrinology & metabolism : JPEM, 26(3-4), 301-308 (2013-01-19)
Congenital hyperinsulinism (CHI) is the most common cause of persistent hypoglycemia in infants. Its management can be extremely complicated, and may involve medical therapy and surgery. The mainstay of the treatment is to maintain normoglycemia, since hypoglycemia during infancy can
Ritika R Kapoor et al.
European journal of endocrinology, 168(4), 557-564 (2013-01-25)
Congenital hyperinsulinism (CHI) is a clinically heterogeneous condition. Mutations in eight genes (ABCC8, KCNJ11, GLUD1, GCK, HADH, SLC16A1, HNF4A and HNF1A) are known to cause CHI. To characterise the clinical and molecular aspects of a large cohort of patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service