Skip to Content
Merck
All Photos(1)

Key Documents

C2174005

Cilazapril impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(1S,9S)-9-{[(R)-1-Ethoxycarbonyl-3-phenylpropyl]amino}-10- oxo-octahydro-6H-pyridazino[1,2a][1,2]diazepine-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31N3O5
CAS Number:
Molecular Weight:
417.50
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cilazapril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cilazapril impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksander Skoczylas et al.
Endokrynologia Polska, 61(1), 21-27 (2010-03-06)
Ghrelin is a polypeptide hormone secreted mainly by the stomach cells, stimulating food intake and growth hormone release. Decreased plasma ghrelin concentration was found in obese subjects. Clinical and experimental data suggest that ghrelin also exerts a blood pressure lowering
Spironolactone attenuates oxidative stress in patients with chronic kidney disease.
Marcin Renke et al.
Hypertension (Dallas, Tex. : 1979), 52(5), e132-e133 (2008-10-01)
Izzet Hosgor et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 19(1), 101-105 (2008-01-09)
Impaired fibrinolysis is associated with thromboembolic complications in hypertensive patients. Cardiovascular morbidity/mortality rates have been reported high even after lowering the elevated blood pressure with antihypertensive drugs. We investigated the effects of clinically used dosages of cilazapril on the fibrinolytic
Milan R Radovanović et al.
Vojnosanitetski pregled, 66(11), 904-908 (2009-12-19)
Poisoning caused by drugs with cardiodepressive effects is an urgent condition in medicine which is associated with high mortality rate regardless of modern therapeutic methods. Accidental or intentional poisoning whit these drugs produces heart activity depression and cardiovascular collapse as
Shih-Hsien Sung et al.
Journal of the Chinese Medical Association : JCMA, 71(5), 247-253 (2008-05-21)
Combined regimen may be superior to monotherapy in blood pressure (BP) control. Since BP control is critically related to cardiovascular mortality and morbidity in hypertensive patients, this study aimed to evaluate the efficacy and safety of a low-dose combined regimen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service