Skip to Content
Merck
All Photos(1)

Key Documents

C1540000

Chlorhexidine dihydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide), Lisium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30Cl2N10 · 2HCl
CAS Number:
Molecular Weight:
578.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorhexidine

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1

InChI

1S/C22H30Cl2N10.2ClH/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;;/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H

InChI key

WJLVQTJZDCGNJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorhexidine dihydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daily chlorhexidine bathing and hospital-acquired infection.
Jean-Jacques Parienti et al.
The New England journal of medicine, 368(24), 2331-2331 (2013-06-14)
Daily chlorhexidine bathing and hospital-acquired infection.
Michael W Climo et al.
The New England journal of medicine, 368(24), 2332-2332 (2013-06-14)
Maristela Dutra-Correa et al.
The journal of adhesive dentistry, 15(3), 287-292 (2013-04-06)
This study tested the hypothesis that 2% chlorhexidine (CHX) does not affect the 18-month clinical performance of two dentin adhesives, XP Bond (XPB) and XENO V (XEN). A total of 120 noncarious cervical lesions (NCCL) were selected and assigned to
Patricia Conley et al.
Nursing, 43(7), 18-23 (2013-06-20)
Several studies have demonstrated that oral care with chlorhexidine gluconate (CHG) 0.12% solution reduces the incidence of ventilator-associated pneumonia (VAP) in mechanically ventilated patients with endotracheal tubes in the ICU. Minimal evidence shows the effectiveness of any oral care protocols
Daily chlorhexidine bathing and hospital-acquired infection.
Anthony L Esposito
The New England journal of medicine, 368(24), 2330-2330 (2013-06-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service