Skip to Content
Merck
All Photos(1)

Key Documents

C1425000

Chloramphenicol sodium succinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Chloramphenicol succinate sodium salt, Chloramphenicol α-succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H15Cl2N2O8Na
CAS Number:
Molecular Weight:
445.18
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chloramphenicol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na].O[C@@H]([C@@H](COC(=O)CCC(O)=O)NC(=O)C(Cl)Cl)c1ccc(cc1)N(=O)=O

InChI

1S/C15H16Cl2N2O8.Na.H/c16-14(17)15(24)18-10(7-27-12(22)6-5-11(20)21)13(23)8-1-3-9(4-2-8)19(25)26;;/h1-4,10,13-14,23H,5-7H2,(H,18,24)(H,20,21);;/t10-,13-;;/m1../s1

InChI key

RJOAHMNSYANTPN-OWVUFADGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chloramphenicol sodium succinate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Ambekar et al.
European journal of clinical pharmacology, 56(5), 405-409 (2000-09-29)
The metabolism of chloramphenicol succinate (CAPS) by human bone marrow was studied in vitro using 75 marrow samples. Whole marrow samples were incubated with CAPS with or without reduced nicotinamide adenine dinucleotide phosphate for 1, 2 and 3 h at
J A Turton et al.
International journal of experimental pathology, 83(5), 225-238 (2003-03-19)
In man, chloramphenicol (CAP), induces two major haemotoxic effects. First, a reversible, dose-related reticulocytopenia and anaemia developing during treatment. Second, a non-dose-related aplastic anaemia (AA), developing weeks after treatment, is often irreversible and fatal. In previous studies, we developed a
M F Festing et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(4), 375-383 (2001-04-11)
Much toxicological research continues to be done using genetically undefined "outbred" stocks of mice and rats, although the case for using isogenic strains has been made repeatedly in the literature over a period of more than two decades. Also, very
Roberta A Gottlieb et al.
Autophagy, 7(4), 434-435 (2010-12-29)
Interventions that reduce infarct size in animal models have largely failed to improve outcome in patients suffering acute myocardial infarction (MI), or 'heart attack'. Our group recently reported a reduction of infarct size by chloramphenicol treatment in a porcine in
N J White et al.
Lancet (London, England), 2(8665), 697-701 (1989-09-23)
An open randomised trial was conducted to compare ceftazidime (120 mg/kg/day) with "conventional therapy" (chloramphenicol 100 mg/kg/day, doxycycline 4 mg/kg/day, trimethoprim 10 mg/kg/day, and sulphamethoxazole 50 mg/kg/day) in the treatment of severe melioidosis. A paired restricted sequential trial designed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service