Skip to Content
Merck
All Photos(1)

Documents

BP617

Aspirin

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Acetylsalicylic acid, 2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aspirin

form

crystalline

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

134-136 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aspirin BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Aspirin Tablets
  • Aspirin Gastro-resistant Tablets
  • Co-codaprin Tablets
  • Aspirin Dispersible Tablets
  • Aspirin and Caffeine Tablets
  • Biochem/physiol Actions

    Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Precautionary Statements

    Hazard Classifications

    Acute Tox. 4 Oral

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 1

    Flash Point(F)

    482.0 °F

    Flash Point(C)

    250 °C


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Validated ultra HPLC method for the simultaneous determination of atorvastatin, aspirin, and their degradation products in capsules.
    Vora D N, et al.
    Journal of Liquid Chromatography and Related Technologies, 31(18), 2821-2837 (2008)
    RP-HPLC analysis of aspirin and clopidogrel bisulphate in combination.
    An K, et al.
    Indian Journal of Pharmaceutical Sciences, 69(4), 597-597 (2007)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service