BCR337
Dibenzo[b,d]furan
BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H8O
CAS Number:
Molecular Weight:
168.19
Beilstein:
121100
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
neat
storage temp.
2-8°C
SMILES string
c1ccc2c(c1)oc3ccccc23
InChI
1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H
InChI key
TXCDCPKCNAJMEE-UHFFFAOYSA-N
Analysis Note
For more information please see:
BCR337
BCR337
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
266.0 °F - closed cup
Flash Point(C)
130 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Juan A Conesa et al.
Chemosphere, 91(2), 118-123 (2012-12-13)
Combustion runs at 700 °C in a horizontal laboratory furnace were carried out on two different electric wires (PVC and halogen-free wire). Tests were performed in the presence and in the absence of the metal conductor of the wires. The
Ruud P M Dings et al.
The Journal of pharmacology and experimental therapeutics, 344(3), 589-599 (2012-12-13)
Galectin-1 (gal-1), which binds β-galactoside groups on various cell surface receptors, is crucial to cell adhesion and migration, and is found to be elevated in several cancers. Previously, we reported on 6DBF7, a dibenzofuran (DBF)-based peptidomimetic of the gal-1 antagonist
Jürgen Wittsiepe et al.
Journal of toxicology and environmental health. Part A, 75(8-10), 423-428 (2012-06-13)
Levels of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/F) were determined in 23 blood samples of exposed workers from a transformer recycling plant in Dortmund, Germany. Compared to the most recent studies on German background levels of adults, elevated concentrations of up
The effect of the substitution position of dibenzofuran on the photophysical and charge-transport properties of host materials for phosphorescent organic light-emitting diodes.
Chil Won Lee et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(4), 1194-1198 (2012-12-21)
Jannie Christensen et al.
Chemistry, an Asian journal, 8(3), 648-652 (2012-12-15)
An intramolecular, organocatalyzed Michael addition has been developed to obtain biologically important 2,3-disubstituted cis-2,3-dihydrobenzofurans. By using mandelic acid salts of primary aminocatalysts, derived from cinchona alkaloids, the intramolecular cyclization reaction has been developed to proceed in high yield, with moderate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service