Skip to Content
Merck
All Photos(1)

Key Documents

B0305000

Beclomethasone dipropionate

anhydrous, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

beclometasone

quality

anhydrous

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

InChI

1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1

InChI key

KUVIULQEHSCUHY-XYWKZLDCSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Beclomethasone dipropionate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Disclaimer

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Adrenal gland,Immune system,Bone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linjie Zhang et al.
Respirology (Carlton, Vic.), 18(2), 272-277 (2012-10-09)
Recent studies have raised concerns about the link between use of inhaled corticosteroids (ICS) and risk of pneumonia in patients with chronic obstructive pulmonary disease. This cross-sectional study aimed to investigate the association between ICS and oropharyngeal colonization by Streptococcus
Eli O Meltzer et al.
Allergy and asthma proceedings, 33(3), 249-257 (2012-06-28)
Intranasal corticosteroids are recommended as first-line therapy for the treatment of the symptoms of persistent allergic rhinitis (AR). Since the phase-out of chlorofluorocarbon nasal aerosols, intranasal corticosteroids have been available only as aqueous nasal sprays. This study was designed to
William J Anderson et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 109(3), 185-189 (2012-08-28)
Whether small airways dysfunction persists in patients with asthma receiving standard community treatment is unknown. Impulse oscillometry (IOS) is a sensitive measure of small airways function. To assess the degree of small airways dysfunction in a cross-section of patients with
Kai-Michael Beeh et al.
The Journal of allergy and clinical immunology, 131(3), 866-874 (2013-02-07)
Allergen-specific TH2 responses contribute to the development of allergic asthma. Their increase may be due to a reduced early exposure to environmental pathogens, which induces a TH1 response, and thereby suppresses the allergic TH2 response. QbG10 (bacteriophage Qbeta-derived virus-like particle
Alberto Papi et al.
Respiratory research, 13, 54-54 (2012-06-27)
Asthma guidelines suggest that therapy can be reduced once asthma is controlled. Despite these recommendations, asthmatic patients are seldom stepped down in clinical practice, and questions remain about when and how to reduce asthma therapy. The purpose of the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service