Skip to Content
Merck
All Photos(1)

Key Documents

ARK2163A

SAFC

Benzyl Alcohol

API grade, Ph. Eur., JP, USP

Pharma Manufacturing

Synonym(s):

Benzyl alcohol, Benzenemethanol, Phenylcarbinol, Phenylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
51241802
E Number:
E1519
EC Index Number:
202-859-9
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

GMP

Agency

JP
Ph. Eur.
USP

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

form

liquid

autoignition temp.

817 °F

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

semi-solid formulation
solid formulation

storage temp.

2-8°C

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Searching for an API that’s right for you? Our high-quality APIs provide all the necessary regulatory information for your needs. We offer best-practice support throughout your entire registration process and boast an excellent track record facilitating our customers’ compliance with international standards – making us your preferred global partner for APIs.
Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Packaging

ARK2163A-1l / 1 l / HDPE bottle
ARK2163A-5l / 5l / HDPE drum
ARK2163A-20l / 20l / HDPE drum
ARK2163A-25l / 25l / HDPE drum

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service