Skip to Content
Merck
All Photos(3)

Key Documents

A0576

Sigma-Aldrich

Agarose

Low EEO

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan, Agarose LE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41105317
PubChem Substance ID:
NACRES:
NA.21

Quality Level

type

Type I-B

form

powder

technique(s)

electrophoresis: suitable

impurities

≤7% water

ash

≤0.25%

turbidity

≤4 NTU (1.5% gel)

EEO

≤0.12

mp

86.5-89.5  °C

transition temp

gel point 36 °C ±1.5 °C (1.5% gel)

gel strength

≥1800 g/cm2 (1% gel)
≥3200 g/cm2 (1.5% gel)

anion traces

sulfate (SO42-): ≤0.12%

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Agarose, a natural polymer is obtained from sea algae. This neutral linear polysaccharide is made up of d-galactose and 3,6-anhydro-l-galactose. These are connected alternatively with the help of glycosidic bonds. Agarose has high significance in gel electrophoresis and gel chromatography.

Application

Agarose has been used:
  • to form the photosynthetic slab
  • in comparative study, to compare it with native agar, and agarose of Gracilaria dura
  • to estimate the amino acid concentrations in agarose and agar samples
  • as immobilisation substrates for enzyme immobilisation
  • for separating high molecular weight nucleic acids at low gel concentrations

Biochem/physiol Actions

Agarose is used as bioink due because of its characteristic properties such as, biocompatibility, mechanical strength and gelling ability at low temperature.

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3D bioprinting cartilage
3D Bioprinting for Reconstructive Surgery, 277-304 (2018)
The `free-flow?biosensor in enzyme inhibition assay
Hatz S and Hall EAH
Sensors and Actuators B, Chemical, 113(2), 700-710 (2006)
Peripheral Nerve Regeneration
Principles of Regenerative Medicine, 1047-1062 (2011)
Oxygen supply by photosynthesis to an implantable islet cell device
Evron Y, et al.
Hormone and Metabolic Research, 47(01), 24-30 (2015)
RECOMBINANT DNA CLONING
Molecular Biology, 45-134 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service