Skip to Content
Merck
All Photos(1)

Key Documents

A0070000

Acesulfame potassium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acesulfame K, 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4KNO4S
CAS Number:
Molecular Weight:
201.24
Beilstein:
3637857
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acesulfame

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[K+].CC1=CC(=O)[N-]S(=O)(=O)O1

InChI

1S/C4H5NO4S.K/c1-3-2-4(6)5-10(7,8)9-3;/h2H,1H3,(H,5,6);/q;+1/p-1

InChI key

WBZFUFAFFUEMEI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acesulfame potassium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Li Li et al.
Journal of molecular neuroscience : MN, 51(1), 225-236 (2013-04-23)
The sweet taste is of immense interest to scientists and has been intensively studied during the last two decades. However, the sweet preference modification and the related mechanisms are still unclear. In this study, we try to establish a mice
Mi-Sun Ha et al.
International journal of food sciences and nutrition, 64(6), 715-723 (2013-05-02)
Using a stepwise assessment of the exposure of Korean consumers to acesulfame K and sucralose, theoretical maximum daily intakes of the sweeteners were calculated using the Budget screening method, which resulted in values greater than the acceptable daily intakes (ADIs).
In search of a role for carbonation: is this a good or bad taste?
Catia Sternini
Gastroenterology, 145(3), 500-503 (2013-07-31)
Pornrat Sinchaipanit et al.
Journal of the science of food and agriculture, 93(13), 3304-3311 (2013-04-16)
Different ratios of combined sweeteners were modified to produce an acceptable reduced-calorie carrot juice. Various hydrocolloids were investigated to improve juice cloud stability. Changes in juice quality attributes were analysed. A combination of the sweeteners aspartame (ASP), acesulfame potassium (ACE)
Francesco Di Salle et al.
Gastroenterology, 145(3), 537-539 (2013-05-30)
Little is known about how CO2 affects neural processing of taste. We used functional magnetic resonance imaging to investigate the effects of carbonation on brain processing of sweet stimuli, which has relevance to studies of food selection and satiety. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service