Skip to Content
Merck
All Photos(1)

Key Documents

939048

Sigma-Aldrich

Lithium bis(trimethylsilyl)amide ChemBeads

Synonym(s):

Hexamethyldisilazane lithium salt ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiC6H18NSi2
Molecular Weight:
167.33
MDL number:
UNSPSC Code:
12352100

description

Organic Salt

Quality Level

form

solid

composition

14-16 wt% loading of base

reaction suitability

core: lithium

SMILES string

[Li]N([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C6H18NSi2.Li/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

YNESATAKKCNGOF-UHFFFAOYSA-N

General description

Lithium bis(trimethylsilyl)amide is a non-nucleophilic strong Brønsted base, which is generally soluble in most of the nonpolar organic solvents. It is most commonly employed in organic reactions.

Application

Base employed in generating enolates for the preparation of lactone precursors, pyranones, and cyclohexanes. Used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines.[6] Also used in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. For general uses, product is also available in powdered form (324620)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkali-metal-catalyzed addition of primary and secondary phosphines to carbodiimides. A general and efficient route to substituted phosphaguanidines
Zhang W X, et al.
Chemical Communications (Cambridge, England), 3812-4, 36-36 (2006)
High-Throughput Reaction Screening with Nanomoles of Solid Reagents Coated on Glass Beads
Tu N P, et al.
Angewandte Chemie (International Edition in English), 58(24), 7987-7991 (2019)
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand
Aguirre A L, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 27(51), 12981-12986 (2021)
Structural Studies of Cesium, Lithium/Cesium, and Sodium/Cesium Bis(trimethylsilyl)amide (HMDS) Complexes
Ojeda-Amador A I, et al.
Inorganic Chemistry, 55(11), 5719-5728 (2016)
Tatsuya Nitabaru et al.
Journal of the American Chemical Society, 131(38), 13860-13869 (2009-09-10)
Full details of an anti-selective catalytic asymmetric nitroaldol reaction promoted by a heterobimetallic catalyst comprised of Nd(5)O(O(i)Pr)(13), an amide-based ligand, and NaHMDS (sodium hexamethyldisilazide) are described. A systematic synthesis and evaluation of amide-based ligands led to the identification of optimum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service