Skip to Content
Merck
All Photos(1)

Key Documents

93535

Supelco

N-(Propionyloxy)succinimide

for LC-MS derivatization, LiChropur, ≥95% (HPLC)

Synonym(s):

1-(1-Oxopropoxy)-2,5-pyrrolidinedione, 2,5-Dioxopyrrolidin-1-yl-propanoate, Propanoic acid-NHS ester, Propionyl-Osu, Succinimidyl propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO4
CAS Number:
Molecular Weight:
171.15
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

quality

LiChropur
for LC-MS derivatization

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

LC/MS: suitable

SMILES string

O=C(CCC1=O)N1OC(CC)=O

InChI

1S/C7H9NO4/c1-2-7(11)12-8-5(9)3-4-6(8)10/h2-4H2,1H3

InChI key

AASBXERNXVFUEJ-UHFFFAOYSA-N

General description

N-(Propionyloxy)succinimide is commonly used as a mobile phase additive in liquid chromatography-mass spectrometry (LC-MS) and its typical application involves the derivatization of histones. It involves the following properties:        
  • Extensive QC testing ensures the highest specifications.        
  • Optimized to improve ionization and resolution.        
  • Extremely low levels of inorganic and organic impurities.        
  • Manufactured specifically for accurate and fast LC-MS.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yea Lu Tay et al.
Scientific reports, 9(1), 19757-19757 (2019-12-26)
Mitragyna speciosa Korth (M. speciosa) has been widely used as a recreational product, however, there are growing concerns on the abuse potentials and toxicity of the plant. Several poisoning and fatal cases involving kratom and mitragynine have been reported but
Somasekhar R Chinnadayyala et al.
Sensors (Basel, Switzerland), 19(24) (2020-01-01)
An electrochemical capacitance immunosensor based on an interdigitated wave-shaped micro electrode array (IDWµE) for direct and label-free detection of C-reactive protein (CRP) was reported. A self-assembled monolayer (SAM) of dithiobis (succinimidyl propionate) (DTSP) was used to modify the electrode array
Zhenfeng Zhang et al.
mBio, 11(3) (2020-06-11)
Archaeal chromatin proteins Cren7 and Sul7d from Sulfolobus are DNA benders. To better understand their architectural roles in chromosomal DNA organization, we analyzed DNA compaction by Cren7 and Sis7d, a Sul7d family member, from Sulfolobus islandicus at the single-molecule (SM)
Yoko Inai et al.
Biochimica et biophysica acta. General subjects, 1864(8), 129632-129632 (2020-05-18)
Mindin (spondin2), a secretory protein related to neural development and immunity, is a member of thrombospondin type I repeat (TSR) superfamily proteins, and has a unique glycosylation of C-mannosylation in its structure. However, it remains unclear whether C-mannosylation plays a
Masaru Fujimoto et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(40), 25150-25158 (2020-09-25)
The plasma membrane (PM) acts as the interface between intra- and extracellular environments and exhibits a tightly regulated molecular composition. The composition and amount of PM proteins are regulated by balancing endocytic and exocytic trafficking in a cargo-specific manner, according

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service